Merck
Environmental enrichment reverses histone methylation changes in the aged hippocampus and restores age-related memory deficits.
Biology (2015-04-04)
Sarah J Morse, Anderson A Butler, Robin L Davis, Ian J Soller, Farah D Lubin
ABSTRAKT
A decline in long-term memory (LTM) formation is a common feature of the normal aging process, which corresponds with abnormal expression of memory-related genes in the aged hippocampus. Epigenetic modulation of chromatin structure is required for proper transcriptional control of genes, such as the brain-derived neurotrophic factor (Bdnf) and Zif268 in the hippocampus during the consolidation of new memories. Recently, the view has emerged that aberrant transcriptional regulation of memory-related genes may be reflective of an altered epigenetic landscape within the aged hippocampus, resulting in memory deficits with aging. Here, we found that baseline resting levels for tri-methylation of histone H3 at lysine 4 (H3K4me3) and acetylation of histone H3 at lysine 9 and 14 (H3K9,K14ac) were altered in the aged hippocampus as compared to levels in the hippocampus of young adult rats. Interestingly, object learning failed to increase activity-dependent H3K4me3 and di-methylation of histone H3 at lysine 9 (H3K9me2) levels in the hippocampus of aged adults as compared to young adults. Treatment with the LSD-1 histone demethylase inhibitor, t-PCP, increased baseline resting H3K4me3 and H3K9,K14ac levels in the young adult hippocampus, while young adult rats exhibited similar memory deficits as observed in aged rats. After environmental enrichment (EE), we found that object learning induced increases in H3K4me3 levels around the Bdnf, but not the Zif268, gene region in the aged hippocampus and rescued memory deficits in aged adults. Collectively, these results suggest that histone lysine methylation levels are abnormally regulated in the aged hippocampus and identify histone lysine methylation as a transcriptional mechanism by which EE may serve to restore memory formation with aging.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride, 97%
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet