Merck
  • Aerobic physical training increases contractile response and reduces cardiac fibrosis in rats subjected to early ovarian hormone deprivation.
Aerobic physical training increases contractile response and reduces cardiac fibrosis in rats subjected to early ovarian hormone deprivation.
Journal of applied physiology (Bethesda, Md. : 1985) (2015-03-21)
Ana Carolina S Felix, Sabrina G V Dutra, Geisa C S V Tezini, Marcus Vinicius Simões, Hugo Celso Dutra de Souza
ABSTRAKT
We investigated the effects of early ovarian hormone deprivation on the heart and the role of physical training in this condition using different approaches: cardiac autonomic tone, contractility, morphology and function, and cardiac fibrosis. Female Wistar rats (n = 48) were assigned into two groups: ovariectomized (Ovx; 10-wk-old) and control rats (Sham; 10-wk-old). Each group was further divided into two subgroups, sedentary and trained (aerobic training by swimming for 10 wk). The sedentary groups showed similar cardiac autonomic tone values; however, only the Sham group had an increase in vagal participation for the determination of the basal heart rate after physical training. The contractile responses to cardiac β-agonists of the sedentary groups were similar, including an increased response to a β1-agonist (dobutamine) observed after physical training. The Ovx sedentary group presented changes in cardiac morphology, which resulted in decreases in the ejection fraction, fractional shortening, and cardiac index compared with the Sham sedentary group. Physical training did little to alter these findings. Moreover, histology analysis showed a significant increase in cardiac fibrosis in the sedentary Ovx group, which was not observed in the trained Ovx group. We conclude that early ovarian hormone deprivation in rats impairs autonomic control, cardiac morphology, and cardiac function and increases cardiac fibrosis; however, it does not affect the contractility induced by dobutamine and salbutamol. Furthermore, this model of physical training prevented an increase in fibrosis and promoted an increase in the cardiac contractile response but had little effect on cardiac autonomic control or morphological and functional parameters.
MATERIAŁY
Numer produktu
Marka
Opis produktu
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%