Merck
  • Odontoblastic Differentiation, Inflammatory Response, and Angiogenic Potential of 4 Calcium Silicate-based Cements: Micromega MTA, ProRoot MTA, RetroMTA, and Experimental Calcium Silicate Cement.
Odontoblastic Differentiation, Inflammatory Response, and Angiogenic Potential of 4 Calcium Silicate-based Cements: Micromega MTA, ProRoot MTA, RetroMTA, and Experimental Calcium Silicate Cement.
Journal of endodontics (2015-06-22)
Seok-Woo Chang, Won-Jung Bae, Jin-Kyu Yi, Soojung Lee, Deok-Won Lee, Kee-Yeon Kum, Eun-Cheol Kim
ABSTRAKT
The aim of this study was to analyze the effects of different calcium silicate-based cements (CSCs) for pulp capping materials including MicroMega MTA (MMTA; MicroMega, Besanchon, France), RetroMTA (RMTA; BioMTA, Seoul, Korea), ProRoot MTA (PMTA; Dentsply, Tulsa, OK), and experimental CSC (ECSC) on odontoblastic differentiation, in vitro angiogenesis, and the inflammatory response in human dental pulp cells. Differentiation was evaluated by alkaline phosphatase activity, alizarin red staining, and reverse-transcriptase polymerase chain reaction (RT-PCR) for the marker genes. The levels of inflammatory mediators and cytokines were measured by RT-PCR and an enzyme-linked immunosorbent assay. In vitro angiogenesis was assessed by RT-PCR for angiogenic genes and an endothelial tube formation assay. PMTA, MMTA, and ECSC increased the alkaline phosphatase activity and mineralization nodule formation and up-regulated messenger RNA (mRNA) expression of odontoblastic markers compared with RMTA. In addition, PMTA, MMTA, and ECSC up-regulated the mRNA of angiogenic genes in human dental pulp cells and increased the capillary tube formation of endothelial cells compared with RMTA. However, all CSCs showed similar expression levels of inducible nitric oxide synthase and cyclooxygenase-2 protein as well as proinflammatory mediators such as nitric oxide, prostaglandin E2, tumor necrosis factor alpha, interleukin (IL)-1β, IL-6, and IL-8 mRNA. Taken together, our experimental results suggest that all CSCs are favorable materials for pulp capping, but PMTA, MMTA, and ECSC may be recommended over RMTA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets