Merck
  • Candida-streptococcal mucosal biofilms display distinct structural and virulence characteristics depending on growth conditions and hyphal morphotypes.
Candida-streptococcal mucosal biofilms display distinct structural and virulence characteristics depending on growth conditions and hyphal morphotypes.
Molecular oral microbiology (2015-03-11)
M M Bertolini, H Xu, T Sobue, C J Nobile, A A Del Bel Cury, A Dongari-Bagtzoglou
ABSTRAKT
Candida albicans and streptococci of the mitis group form communities in multiple oral sites, where moisture and nutrient availability can change spatially or temporally. This study evaluated structural and virulence characteristics of Candida-streptococcal biofilms formed on moist or semidry mucosal surfaces, and tested the effects of nutrient availability and hyphal morphotype on dual-species biofilms. Three-dimensional models of the oral mucosa formed by immortalized keratinocytes on a fibroblast-embedded collagenous matrix were used. Infections were carried out using Streptococcus oralis strain 34, in combination with a C. albicans wild-type strain, or pseudohyphal-forming mutant strains. Increased moisture promoted a homogeneous surface biofilm by C. albicans. Dual biofilms had a stratified structure, with streptococci growing in close contact with the mucosa and fungi growing on the bacterial surface. Under semidry conditions, Candida formed localized foci of dense growth, which promoted focal growth of streptococci in mixed biofilms. Candida biofilm biovolume was greater under moist conditions, albeit with minimal tissue invasion, compared with semidry conditions. Supplementing the infection medium with nutrients under semidry conditions intensified growth, biofilm biovolume and tissue invasion/damage, without changing biofilm structure. Under these conditions, the pseudohyphal mutants and S. oralis formed defective superficial biofilms, with most bacteria in contact with the epithelial surface, below a pseudohyphal mass, resembling biofilms growing in a moist environment. The presence of S. oralis promoted fungal invasion and tissue damage under all conditions. We conclude that moisture, nutrient availability, hyphal morphotype and the presence of commensal bacteria influence the architecture and virulence characteristics of mucosal fungal biofilms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hematoxylin
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Hematoxylin, certified by the BSC
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Informacje o cenach i dostępności nie są obecnie dostępne.