MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Xenosiderophore Utilization Promotes Bacteroides thetaiotaomicron Resilience during Colitis.
Xenosiderophore Utilization Promotes Bacteroides thetaiotaomicron Resilience during Colitis.
Cell host & microbe (2020-02-23)
Wenhan Zhu, Maria G Winter, Luisella Spiga, Elizabeth R Hughes, Rachael Chanin, Aditi Mulgaonkar, Jenelle Pennington, Michelle Maas, Cassie L Behrendt, Jiwoong Kim, Xiankai Sun, Daniel P Beiting, Lora V Hooper, Sebastian E Winter
ABSTRACT
During short-lived perturbations, such as inflammation, the gut microbiota exhibits resilience and reverts to its original configuration. Although microbial access to the micronutrient iron is decreased during colitis, pathogens can scavenge iron by using siderophores. How commensal bacteria acquire iron during gut inflammation is incompletely understood. Curiously, the human commensal Bacteroides thetaiotaomicron does not produce siderophores but grows under iron-limiting conditions using enterobacterial siderophores. Using RNA-seq, we identify B. thetaiotaomicron genes that were upregulated during Salmonella-induced gut inflammation and were predicted to be involved in iron uptake. Mutants in the xusABC locus (BT2063-2065) were defective for xenosiderophore-mediated iron uptake in vitro. In the normal mouse gut, the XusABC system was dispensable, while a xusA mutant colonized poorly during colitis. This work identifies xenosiderophore utilization as a critical mechanism for B. thetaiotaomicron to sustain colonization during inflammation and suggests a mechanism of how interphylum iron metabolism contributes to gut microbiota resilience.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, Suitable for determination of Fe
Sigma-Aldrich
Ammonium iron(III) citrate, reagent grade, powder
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Protoporphyrin IX, ≥95%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Enterobactin, from Escherichia coli, ≥98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Calcium chloride dihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Cobalt(II) chloride hexahydrate, reagent grade
Sigma-Aldrich
Neomycin trisulfate salt hydrate, powder
Supelco
Metronidazole, analytical standard