MilliporeSigma
  • [L-carnitine treatment and fish odor syndrome: an unwaited adverse effect].
[L-carnitine treatment and fish odor syndrome: an unwaited adverse effect].
Revue neurologique (2011-04-13)
F Rocher, C Caruba, F Broly, C Lebrun
ABSTRACT
Levocarnitine treatment is usually well tolerated, with essentially dose-dependent diarrhea as the main induced adverse effect. We report a case of fish odor syndrome during levocarnitine treatment which resolved after levocarnitine discontinuation. This adverse effect seems to be correlated with excedent carnitine intake and might be expressed when the elimination pathway becomes saturated or in a situation of deficiency enzymatic metabolism.
Sigma-Aldrich
N,N-Dimethylglycine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
N,N-Dimethylglycine hydrochloride
Pricing and availability is not currently available.