MilliporeSigma
  • A conductance study of guanidinium chloride, thiocyanate, sulfate, and carbonate in dilute aqueous solutions: ion-association and carbonate hydrolysis effects.
A conductance study of guanidinium chloride, thiocyanate, sulfate, and carbonate in dilute aqueous solutions: ion-association and carbonate hydrolysis effects.
The journal of physical chemistry. B (2012-12-18)
Johannes Hunger, Roland Neueder, Richard Buchner, Alexander Apelblat
ABSTRACT
We study the conductance of dilute aqueous solutions for a series of guandinium salts at 298.15 K. The experimental molar conductivities were analyzed within the framework of the Quint-Viallard theory in combination with Debye-Hückel activity coefficients. From this analysis, we find no evidence for significant ion association in aqueous solutions of guanidinium chloride (GdmCl) and guanidinium thiocyanate (GdmSCN), and the molar conductivity of these electrolytes can be modeled assuming a complete dissociation. The limiting ionic conductivity of the guanidinium ion (Gdm(+)) is accurately determined to λ(Gdm(+)) = 51.45 ± 0.10 S cm(2) mol(-1). For the bivalent salts guanidinium sulfate (Gdm(2)SO(4)) and guanidinium carbonate (Gdm(2)CO(3)), the molar conductivities show small deviations from ideal (fully dissociated electrolyte) behavior, which are related to weak ion association in solution. Furthermore, for solutions of Gdm(2)CO(3), the hydrolysis of the carbonate anion leads to distinctively increased molar conductivities at high dilutions. The observed ion association is rather weak for all studied electrolytes and cannot explain the different protein denaturing activities of the studied guanidinium salts, as has been proposed previously.
Sigma-Aldrich
Guanidine thiocyanate, BioReagent, Molecular Biology, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Guanidine thiocyanate, Molecular Biology, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Guanidine thiocyanate solution, BioUltra, Molecular Biology, ~6 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Guanidine thiocyanate, BioUltra, Molecular Biology, ≥99.0% (AT)
Pricing and availability is not currently available.