MilliporeSigma
Tramadol inhibits the contractility of isolated human myometrium.
Autonomic & autacoid pharmacology (2013-02-26)
N H Shah, E Thomas, R Jose, J Peedicayil
ABSTRACT
This study was conducted to determine whether the atypical opioid analgesic tramadol inhibits the contractility of isolated non-pregnant human myometrium. Ten strips of non-pregnant human myometrium stimulated with 55 mm potassium chloride (KCl) were treated with three concentrations (30, 100 and 300 μm) of tramadol to test for any inhibitory effect of tramadol. The effects of concurrent administration of the ß adrenoceptor antagonist propranolol (1 μm), the guanylyl cyclase and nitric oxide synthase inhibitor methylene blue (20 μm) and the opioid receptor antagonist naloxone (100 μm) with tramadol were also studied. Tramadol caused a concentration-dependent inhibition of KCl-induced myometrial contractility, which was statistically significant at all three concentrations of tramadol used. Propranolol significantly reversed the inhibitory effect of 100 μm tramadol on KCl-induced myometrial contractility but not that of 300 μm tramadol. Neither methylene blue nor naloxone reversed the inhibitory effect of tramadol on KCl-induced myometrial contractility. These results suggest that tramadol inhibits KCl-induced contractility of isolated human myometrium. They also suggest that tramadol relaxes the myometrium due to stimulation of ß1 adrenoceptors. However, the concentrations of tramadol required to relax the myometrium were high and likely to be attained at toxic doses, rather than therapeutic doses, of tramadol.
Sigma-Aldrich
Isoprenaline hydrochloride
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylene Blue solution
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Potassium standard solution, suitable for atomic absorption spectrometry, 1000 ppm K+, 1 mg/mL K+
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl
Supelco
Potassium chloride solution, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride, ReagentPlus®, ≥99.0%