MilliporeSigma
  • Induction of miR-21-PDCD4 signaling by tungsten carbide-cobalt nanoparticles in JB6 cells involves ROS-mediated MAPK pathways.
Induction of miR-21-PDCD4 signaling by tungsten carbide-cobalt nanoparticles in JB6 cells involves ROS-mediated MAPK pathways.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer (2013-06-14)
Lichao Hou, Linda Bowman, Terence G Meighan, Xianglin Shi, Min Ding
ABSTRACT
Tungsten carbide-cobalt (WC-Co) nanoparticle composites have wide applications because of their hardness and toughness. WC-Co was classified as "probably carcinogenic" to humans by the International Agency for Research on Cancer (IARC) in 2003. It is believed that the toxicity and carcinogenesis of WC-Co is associated with particle size. Recent studies demonstrated that the tumor suppressor gene programmed cell death 4 (PDCD4) and its upstream regulator miR-21 have been considered as oncogenes for novel cancer prevention or anticancer therapies. The present study examined the effects of WC-Co nanoparticles on miR-21-PDCD4 signaling in a mouse epidermal cell line (JB6 P+). The results showed that (i) exposure of JB6 cells to WC-Co stimulated a increase of miR-21 generation; (ii) WC-Co also caused inhibition of PDCD4, a tumor suppressor protein and downstream target of miR-21, expression in JB6 cells; (iii) inhibition of ERKs with ERK inhibitor U0126 significantly reversed WC-Cominus;induced PDCD4 inhibition, but inhibition of p38 with p38 inhibitor SB203580 did not; and (iv) ROS scavengers, N-acetyl-L-cysteine and catalase, blocked the inhibitory effect of WC-Co on PDCD4 expression, while superoxide dismutase promoted the inhibitory effect. These findings demonstrate that WC-Co nanoparticles induce miR-21 generation, but inhibit PDCD4 production, which may be mediated through ROS, especially endogenous H2O2, and ERK pathways. Unraveling the complex mechanisms associated with these events may provide insights into the initiation and progression of WC-Co-induced carcinogenesis.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Tungsten(IV) carbide, powder, 2 μm, ≥99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%