MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Phytochemical screening and in-vitro antioxidant activity isolated bioactive compounds from Tridax procumbens Linn.
Phytochemical screening and in-vitro antioxidant activity isolated bioactive compounds from Tridax procumbens Linn.
Pakistan journal of biological sciences : PJBS (2014-02-13)
Manjusha Saxena, Abrar Hussain Mir, Manik Sharma, Mohd Yousu Malla, Sumeerah Qureshi, Mohd Iqbal Mir, Yogesh Chaturvedi
ABSTRACT
Tridax procumbens L., Asteraceae, has been extensively used for various ailments in the Ayurvedic system of medicine. Previous studies have revealed remarkable phytoconstituents from Tridax procumbens L. with significant antioxidant activity. The aim of the present study is to measure the anti-DPPH activity of the purified isolated compounds from n butanol soluble part and ethyl acetate soluble part of successive methanolic extract of Tridax procumbens L. We thus quantified the total phenolic and total flavonoids in different purified isolated compounds, the whole of the tests were evaluated with a sample cone. of 100 microg mL(-1) and were determined spectrophotometrically using Folin-ciocaltue and AlCl3 reagents, respectively. DPPH (1,1-diphenyl, 2-picryl hydrazyl) assay was used to determine the in vitro antioxidant activity of different isolated compounds. Isolated compounds, one from ethyl acetate soluble part (EF-I) and one from n butanol soluble part (BF-II) were reported to possess a significant anti DPPH activity with lowest IC50 values 67.26 and 80.90 microg mL(-1), respectively while comparable to standard ascorbic acid with IC50 value of 59.62 microg mL(-1), due to the high concentration of phenols 146.4 microg mL(-1) from EF-I and 142.2 microg mL(-1) from BF-II and flavonoids 48 and 42.5 microg mL(-1) found in EF-I and BF-II isolated compounds, respectively.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethyl acetate, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethyl acetate, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free