MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Association between hypogonadism, symptom burden, and survival in male patients with advanced cancer.
Association between hypogonadism, symptom burden, and survival in male patients with advanced cancer.
Cancer (2014-03-01)
Rony Dev, David Hui, Egidio Del Fabbro, Marvin O Delgado-Guay, Nikhil Sobti, Shalini Dalal, Eduardo Bruera
ABSTRACT
A high frequency of hypogonadism has been reported in male patients with advanced cancer. The current study was performed to evaluate the association between low testosterone levels, symptom burden, and survival in male patients with cancer. Of 131 consecutive male patients with cancer, 119 (91%) had an endocrine evaluation of total (TT), free (FT), and bioavailable testosterone (BT); high-sensitivity C-reactive protein (CRP); vitamin B12; thyroid-stimulating hormone; 25-hydroxy vitamin D; and cortisol levels when presenting with symptoms of fatigue and/or anorexia-cachexia. Symptoms were evaluated by the Edmonton Symptom Assessment Scale. The authors examined the correlation using the Spearman test and survival with the log-rank test and Cox regression analysis. The median age of the patients was 64 years; the majority of patients were white (85 patients; 71%). The median TT level was 209 ng/dL (normal: ≥ 200 ng/dL), the median FT was 4.4 ng/dL (normal: ≥ 9 ng/dL), and the median BT was 22.0 ng/dL (normal: ≥ 61 ng/dL). Low TT, FT, and BT values were all associated with worse fatigue (P ≤ .04), poor Eastern Cooperative Oncology Group performance status (P ≤ .05), weight loss (P ≤ .01), and opioid use (P ≤ .005). Low TT and FT were associated with increased anxiety (P ≤ .04), a decreased feeling of well-being (P ≤ .04), and increased dyspnea (P ≤ .05), whereas low BT was only found to be associated with anorexia (P = .05). Decreased TT, FT, and BT values were all found to be significantly associated with elevated CRP and low albumin and hemoglobin. On multivariate analysis, decreased survival was associated with low TT (hazards ratio [HR], 1.66; P = .034), declining Eastern Cooperative Oncology Group performance status (HR, 1.55; P = .004), high CRP (HR, 3.28; P < .001), and decreased albumin (HR, 2.52; P < .001). In male patients with cancer, low testosterone levels were associated with systemic inflammation, weight loss, increased symptom burden, and decreased survival. A high frequency of hypogonadism has been reported in male patients with advanced cancer. In the current study, an increased symptom burden, systemic inflammation, weight loss, opioid use, and poor survival were found to be associated with decreased testosterone levels in male patients with cancer. Cancer 2014;120:1586-1593. © 2014 American Cancer Society.
Product Description
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Cyanocobalamin (Vitamin B12) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Testosterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Thyrotropic hormone from bovine pituitary
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Thyrotropic hormone from human pituitary, lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Testosterone, ≥98%
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®