MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Flotillin depletion affects ErbB protein levels in different human breast cancer cells.
Flotillin depletion affects ErbB protein levels in different human breast cancer cells.
Biochimica et biophysica acta (2014-04-22)
Nagham Asp, Sascha Pust, Kirsten Sandvig
ABSTRACT
The ErbB3 receptor is an important regulator of cell growth and carcinogenesis. Among breast cancer patients, up to 50-70% have ErbB3 overexpression and 20-30% show overexpressed or amplified ErbB2. ErbB3 has also been implicated in the development of resistance to several drugs used against cancers driven by ErbB1 or ErbB2. One of the main challenges in ErbB-targeting therapy is to inactivate signaling mediated by ErbB2-ErbB3 oncogenic receptor complexes. We analyzed the regulatory role of flotillins on ErbB3 levels and ErbB2-ErbB3 complexes in SKBR3, MCF7 and MDA-MB-134-VI human breast cancer cells. Recently, we described a mechanism for interfering with ErbB2 signaling in breast cancer and demonstrated a molecular complex of flotillin scaffolding proteins with ErbB2 and Hsp90. In the present study, flotillins were found to be in a molecular complex with ErbB3, even in cells without the presence of ErbB2 or other ErbB receptors. Depletion of either flotillin-1 or flotillin-2 resulted in downregulation of ErbB3 and a selective reduction of ErbB2-ErbB3 receptor complexes. Moreover, flotillin-2 depletion resulted in reduced activation of Akt and MAPK signaling cascades, and as a functional consequence of flotillin depletion, breast cancer cells showed an impaired cell migration. Altogether, we provide data demonstrating a novel and functional role of flotillins in the regulation of ErbB protein levels and stabilization of ErbB2-ErbB3 receptor complexes. Thus, flotillins are crucial regulators for oncogenic ErbB function and potential targets for cancer treatment.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
DAPI, for nucleic acid staining
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Geldanamycin from Streptomyces hygroscopicus, ≥98% (HPLC), powder
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%