MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis, cytotoxicity, and phase-solubility study of cyclodextrin click clusters.
Synthesis, cytotoxicity, and phase-solubility study of cyclodextrin click clusters.
Journal of pharmaceutical sciences (2014-08-22)
Hoa Thi Le, Hyun Mi Jeon, Choon Woo Lim, Tae Woo Kim
ABSTRACT
To explore the possibility of cyclodextrin click clusters (CCCs) as a new cyclodextrin-based excipient, we prepared three different CCCs; heptakis{6-(4-hydroxymethyl-1H-[1,2,3]triazol-1-yl)-6-deoxy}-β-cyclodextrin (HT-β-CD), heptakis{6-(4-hydroxymethyl-1H-[1,2,3]triazol-1-yl)-6-deoxy}{2,3-di-O-methyl}-β-cyclodextrin (HT-β-CD(OMe)2 ), and heptakis{6-(4-sulfonylmethyl-1H-[1,2,3]triazol-1-yl)-6-deoxy}-β-cyclodextrin (ST-β-CD). The CCCs were prepared using copper(I)-catalyzed azide-alkyne cycloaddition from 6-azido-6-deoxy-β-CD and their water solubility, cytotoxicity, and drug-solubilizing effect were investigated. Water turbidity testing of the CCCs showed that the minimum water solubility of the CCCs is at least 20 times higher than that of β-CD. An MTT cell viability assay performed on HeLa cells demonstrated a low cytotoxicity of the CCCs compared with 2,6-dimethyl-β-cyclodextrin. HT-β-CD(OMe)2 and ST-β-CD did not demonstrate any cytotoxicity within the experimental concentration (∼5 mM) like 2-hydroxypropyl-β-CD. A phase-solubility study of prednisolone with the CCCs suggested that CCCs showed increased solubility of prednisolone in the presence of increasing concentrations of the CCCs. The comparison between the conventional CD derivatives and CCCs on solubility, cytotoxicity, and binding property implies that CCCs are alternative cyclodextrin derivatives useful for overcoming the restrictions of conventional cyclodextrin chemistry.
Product Description
Prednisolone, European Pharmacopoeia (EP) Reference Standard
USP
Prednisolone, United States Pharmacopeia (USP) Reference Standard
Supelco
Prednisolone, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisolone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Prednisolone, ≥99%
Supelco
Prednisolone, VETRANAL®, analytical standard
Prednisolone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Prednisolone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Heptakis(2,6-di-O-methyl)-β-cyclodextrin
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin, average Mw ~1,540
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture