MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • DNA damage response and anti-apoptotic proteins predict radiosensitization efficacy of HDAC inhibitors SAHA and LBH589 in patient-derived glioblastoma cells.
DNA damage response and anti-apoptotic proteins predict radiosensitization efficacy of HDAC inhibitors SAHA and LBH589 in patient-derived glioblastoma cells.
Cancer letters (2014-10-12)
Lotte M E Berghauser Pont, Kishan Naipal, Jenneke J Kloezeman, Subramanian Venkatesan, Martin van den Bent, Dik C van Gent, Clemens M F Dirven, Roland Kanaar, Martine L M Lamfers, Sieger Leenstra
ABSTRACT
HDAC inhibitors have radiosensitizing effects in established cancer cell lines. This study was conducted to compare the efficacy of SAHA, LBH589, Valproic Acid (VPA), MS275 and Scriptaid in the patient-derived glioblastoma model. In more detail, SAHA and LBH589 were evaluated to determine predictors of response. Acetylated-histone-H3, γH2AX/53BP1, (p)Chek2/ATM, Bcl-2/Bcl-XL, p21(CIP1/WAF1) and caspase-3/7 were studied in relation to response. SAHA sensitized 50% of cultures, LBH589 45%, VPA and Scriptaid 40% and MS275 60%. Differences after treatment with SAHA/RTx or LBH589/RTx in a sensitive and resistant culture were increased acetylated-H3, caspase-3/7 and prolonged DNA damage repair γH2AX/53BP1 foci. pChek2 was found to be associated with both SAHA/RTx and LBH589/RTx response with a positive predictive value (PPV) of 90%. Bcl-XL had a PPV of 100% for LBH589/RTx response. Incubation with HDACi 24 and 48 hours pre-RTx resulted in the best efficacy of combination treatment. In conclusion a subset of patient-derived glioblastoma cultures were sensitive to HDACi/RTx. For SAHA and LBH589 responses were strongly associated with pChek2 and Bcl-XL, which warrant further clinical exploration. Additional information on responsiveness was obtained by DNA damage response markers and apoptosis related proteins.
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, SAJ first grade, ≥98.0%
Sigma-Aldrich
Acrylamide, ≥99.9%
Sigma-Aldrich
Acrylamide, ≥98.0%