MilliporeSigma
  • FOXF1 mediates mesenchymal stem cell fusion-induced reprogramming of lung cancer cells.
FOXF1 mediates mesenchymal stem cell fusion-induced reprogramming of lung cancer cells.
Oncotarget (2014-09-23)
Hong-Jian Wei, Jac A Nickoloff, Wei-Hong Chen, Hen-Yu Liu, Wen-Cheng Lo, Ya-Ting Chang, Pan-Chyr Yang, Cheng-Wen Wu, David F Williams, Juri G Gelovani, Win-Ping Deng
ABSTRACT
Several reports suggest that malignant cells generate phenotypic diversity through fusion with various types of stromal cells within the tumor microenvironment. Mesenchymal stem cell (MSC) is one of the critical components in the tumor microenvironment and a promising fusogenic candidate, but the underlying functions of MSC fusion with malignant cell have not been fully examined. Here, we demonstrate that MSCs fuse spontaneously with lung cancer cells, and the latter is reprogrammed to slow growth and stem-like state. Transcriptome profiles reveal that lung cancer cells are reprogrammed to a more benign state upon MSC fusion. We further identified FOXF1 as a reprogramming mediator that contributes not only to the reprogramming toward stemness but also to the p21-regulated growth suppression in fusion progeny. Collectively, MSC fusion does not enhance the intrinsic malignancy of lung cancer cells. The anti-malignant effects of MSC fusion-induced reprogramming on lung cancer cells were accomplished by complementation of tumorigenic defects, including restoration of p21 function and normal terminal differentiation pathways as well as up-regulation of FOXF1, a putative tumor suppressor. Such fusion process raises the therapeutic potential that MSC fusion can be utilized to reverse cellular phenotypes in cancer.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Demecolcine, ≥98% (HPLC)
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Supelco
Methanol, analytical standard