MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of solvent on percutaneous absorption of nonvolatile lipophilic solute.
Effects of solvent on percutaneous absorption of nonvolatile lipophilic solute.
International journal of pharmaceutics (2014-09-28)
Rattikorn Intarakumhaeng, S Kevin Li
ABSTRACT
Understanding the effects of solvents upon percutaneous absorption can improve drug delivery across skin and allow better risk assessment of toxic compound exposure. The objective of the present study was to examine the effects of solvents upon the deposition of a moderately lipophilic solute at a low dose in the stratum corneum (SC) that could influence skin absorption of the solute after topical application. Skin permeation experiments were performed using Franz diffusion cells and human epidermal membrane (HEM). Radiolabeled corticosterone ((3)H-CS) was the model permeant. The solvents used had different evaporation and skin penetration properties that were expected to impact skin deposition of CS and its absorption across skin. The results show no correlation between the rate of absorption of the permeant and the rate of solvent evaporation/penetration with ethanol, hexane, isopropanol, and butanol as the solvent; all of these solvents have fast evaporation rates (complete evaporation in <30 min after application). This suggests no differences in solvent-induced deposition of CS in the SC for the fast-evaporating solvents. The results of these fast-evaporating solvents were different from those of water, propylene glycol, and polyethylene glycol 400, that a relationship between permeant absorption and the rate of solvent evaporation was observed.
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%