MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Expression and functions of ASIC1 in the zebrafish retina.
Biochemical and biophysical research communications (2014-12-03)
Sha Liu, Mei-Xia Wang, Cheng-Jie Mao, Xiao-Yu Cheng, Chen-Tao Wang, Jian Huang, Zhao-Min Zhong, Wei-Dong Hu, Fen Wang, Li-Fang Hu, Han Wang, Chun-Feng Liu
ABSTRACT
It has been demonstrated that acid sensing ionic channels (ASICs) are present in the central and peripheral nervous system of mammals, including the retina. However, it remains unclear whether the zebrafish retina also expresses ASICs. In the present study, the expression and distribution of zasic1 were examined in the retina of zebrafish. Both zasic1 mRNA and protein expressions were detected in the adult zebrafish retina. A wide distribution of ASIC1 in zebrafish retina was confirmed using whole mount in situ hybridization and immunohistochemistry study. Acidosis-induced currents in the isolated retinal ganglion cells (RGCs) were also recorded using whole cell patch clamping. Moreover, blockade of ASICs channel significantly reduced the locomotion of larval zebrafish in response to light exposure. In sum, our data demonstrate the presence of ASIC1 and its possible functional relevance in the retina of zebrafish.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%