MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Melatonin protects testes against lithium-pilocarpine-induced temporal lobe epilepsy in rats: a time course study.
Melatonin protects testes against lithium-pilocarpine-induced temporal lobe epilepsy in rats: a time course study.
Andrologia (2014-04-02)
S Shokri, M Kazemi, M A Firouzjaei, M Hemadi, A Moayeri, M Ganjkhani, R Nejatbakhsh
ABSTRACT
Male dysfunction is common in patients with temporal lobe epilepsy (TLE). We evaluated whether melatonin, as a supplement, can play a positive role in reducing the epileptogenesis imposing abnormalities of spermatozoa and testes in epileptic rats. Status epilepticus was induced based on the TLE lithium-pilocarpine model. Two patterns of melatonin were administered to the epileptic animals along the mean durations of latent (14 days) and chronic (60 days) phases. Sperm parameters, different antioxidant enzyme levels, germ cell apoptosis, body and relative sex organ weights were evaluated in all groups 60 days following SE induction. Chronic TLE caused a significant reduction in sperm parameters. In the testis, the reduced level of antioxidant enzymes was accompanied by a significant increase in malondialdehyde concentration. The presence of oxidant condition in the testes of epileptic animals caused expanded apoptosis in the germ cell layer. Moreover, the amount of weight gain in epileptic animals was more prominent. Melatonin administration was able to improve sperm motility by increasing the total antioxidant level. There was also a significant reduction in the spermatogenic cell line apoptosis and the extra weight gain of melatonin-treated animals. Melatonin supplementation might be considered as an acceptable cotreatment in epileptic patients.
Product Description
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Pilocarpine hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%