MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • PCK3145 inhibits proliferation and induces apoptosis in breast and colon cancer cells.
PCK3145 inhibits proliferation and induces apoptosis in breast and colon cancer cells.
Anticancer research (2015-03-10)
Antonios Kostopoulos, Efstathia Papageorgiou, Michael Koutsilieris, Gregory Sivolapenko
ABSTRACT
To explore the effects of PCK3145 beyond prostate cancer. Using Trypan blue, MTT proliferation assays, cell cycle and apoptosis analysis, we assessed the effects of PCK3145 on prostate (PC-3), breast (MCF-7) and colon (HT-29) human cancer cell lines and in osteosarcoma (MG-63) cells; any synergistic effects with docetaxel and oxaliplatin were also explored. PCK3145 inhibited proliferation and induced apoptosis of PC-3, MCF-7 and HT-29 cells in a dose- and time-dependent manner but not in the MG-63 cell line, consistent with the low expression of the laminin receptor (LR) in the latter cell line. PCK3145 produced rapid (within 5 min) and transient (up to 60 min) activation of MEK and ERK1/2. Synergistic effects were observed with docetaxel and oxaliplatin. PCK3145 can exert anticancer activity not only on prostate but also on breast and colon cancer cells, possibly through LR-mediated activation of MEK and ERK1/2 phosphorylation.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M