MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • VEGF-A promotes both pro-angiogenic and neurotrophic capacities for nerve recovery after compressive neuropathy in rats.
VEGF-A promotes both pro-angiogenic and neurotrophic capacities for nerve recovery after compressive neuropathy in rats.
Molecular neurobiology (2014-05-29)
Julien Pelletier, Emilie Roudier, Pierre Abraham, Bérengère Fromy, Jean Louis Saumet, Olivier Birot, Dominique Sigaudo-Roussel
ABSTRACT
Nerve recovery following injury is usually incomplete, leaving functional deficits. Our aim was to investigate the neural changes in pro-angiogenic, pro-inflammatory and apoptotic factors during and after chronic nerve compression (CNC). Nerve function was impaired after CNC and was progressively restored after nerve decompression, while nerve blood flow was elevated. While the expression of the pro-inflammatory and pro-angiogenic cytokines IL-6, TNF-α and VEGF-A was high during and after CNC, we observed that inhibition of VEGF-A receptors strongly counteracted the angiogenic response induced by the ex vivo CNC. Activation of the pro-survival transcription factor nuclear factor-kappa B (NF-κB) increased during CNC, returning to control levels after nerve decompression. After nerve decompression, the downregulation of Mdm2 correlated well with an increased expression of pro-apoptotic transcription factor p53. All together, we bring novel evidence that CNC activates transcription factors such as NF-κB and p53, which are key effectors of the cellular stress response, suggesting a neuroprotective process associated with an increased VEGF-A-mediated neurotrophic effect. Our results highlight the role of pro-angiogenic and pro-inflammatory cytokines during CNC that are reinforced by increasing neurotrophic capacity during recovery to promote nerve regeneration.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%