MilliporeSigma
  • Suppression of TDO-mediated tryptophan catabolism in glioblastoma cells by a steroid-responsive FKBP52-dependent pathway.
Suppression of TDO-mediated tryptophan catabolism in glioblastoma cells by a steroid-responsive FKBP52-dependent pathway.
Glia (2014-08-19)
Martina Ott, Ulrike M Litzenburger, Katharina J Rauschenbach, Lukas Bunse, Katharina Ochs, Felix Sahm, Stefan Pusch, Christiane A Opitz, Jonas Blaes, Andreas von Deimling, Wolfgang Wick, Michael Platten
ABSTRACT
Tryptophan catabolism is increasingly recognized as a key and druggable molecular mechanism active in cancer, immune, and glioneural cells and involved in the modulation of antitumor immunity, autoimmunity and glioneural function. In addition to the pivotal rate limiting enzyme indoleamine-2,3-dioxygenase, expression of tryptophan-2,3-dioxygenase (TDO) has recently been described as an alternative pathway responsible for constitutive tryptophan degradation in malignant gliomas and other types of cancer. In addition, TDO has been implicated as a key regulator of neurotoxicity involved in neurodegenerative diseases and ageing. The pathways regulating TDO expression, however, are largely unknown. Here, a siRNA-based transcription factor profiling in human glioblastoma cells revealed that the expression of human TDO is suppressed by endogenous glucocorticoid signaling. Similarly, treatment of glioblastoma cells with the synthetic glucocorticoid dexamethasone led to a reduction of TDO expression and activity in vitro and in vivo. TDO inhibition was dependent on the immunophilin FKBP52, whose FK1 domain physically interacted with the glucocorticoid receptor as demonstrated by bimolecular fluorescence complementation and in situ proximity ligation assays. Accordingly, gene expression profile analyses revealed negative correlation of FKBP52 and TDO in glial and neural tumors and in normal brain. Knockdown of FKBP52 and treatment with the FK-binding immunosuppressant FK506 enhanced TDO expression and activity in glioblastoma cells. In summary, we identify a novel steroid-responsive FKBP52-dependent pathway suppressing the expression and activity of TDO, a central and rate-limiting enzyme in tryptophan metabolism, in human gliomas.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
USP
Tacrolimus, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Acetone, ≥99.5%, suitable for residue analysis