MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Simultaneous detection of multi-allergens in an incurred food matrix using ELISA, multiplex flow cytometry and liquid chromatography mass spectrometry (LC-MS).
Simultaneous detection of multi-allergens in an incurred food matrix using ELISA, multiplex flow cytometry and liquid chromatography mass spectrometry (LC-MS).
Food chemistry (2015-01-13)
Ahmed Gomaa, Joyce Boye
ABSTRACT
Food allergy is a public health concern and an important food safety issue. Food allergies affect up to 6% of infants and children and 4% of adults. The objective of this work was to determine differences in the detection of single and multiple allergens (i.e., casein, soy protein, and gluten) in an incurred food matrix before and after baking. Cookies were used as a model food system. Three methods, namely, multiplex assay (a new optimized method based on flow cytometry for multiple allergen analysis), enzyme-linked immunosorbent assay (ELISA) using commercial kits and LC-MS were used to detect allergens in the samples before and after baking. The ELISA kits performed well in detecting allergens in the raw samples with recoveries of 91-108%, 88-127% and 85-108% for casein, soy protein and gluten, respectively. Recoveries were poor for the baked cookies (67-90%, 66-95% and 66-88% for casein, soy protein and gluten, respectively). The multiplex flow cytometry assay permitted multiple allergen detection in the raw samples, with the following recoveries based on soluble protein: casein, 95-107%; soy protein, 92-97%, and gluten, 96-99%. Data for the baked cookies were as follows: casein, 84-90%; soy protein, 80-88%, and gluten, 80-90%. The LC-MS technique detected marker peptides that could be used to identify allergens in the baked food samples up to concentrations of 10 ppm for casein and soy protein, and 100 ppm for gluten. To the best of our knowledge, the current study is the first to compare ELISA, LC-MS and multiplex flow cytometry methods for the detection of multiple allergens simultaneously incurred in a model food system.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Supelco
1,2-Dichloroethane, analytical standard
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing