MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • In vitro evaluation of TAT-OP1 osteogenic properties and prospects for in vivo applications.
In vitro evaluation of TAT-OP1 osteogenic properties and prospects for in vivo applications.
Journal of tissue engineering and regenerative medicine (2012-09-14)
R Di Liddo, C Grandi, D Dalzoppo, V Villani, M Venturini, A Negro, L Sartore, M Artico, M T Conconi, P P Parnigotto
ABSTRACT
So far, osteogenic protein 1 (OP1) is biotechnologically produced and approved for the treatment of human lumbar spine fusion and long bone non-union fractures. When combined with the TAT sequence, it has been demonstrated in vitro to be easily taken up by PC12 neuronal cells and to acquire its biological activity after intracellular refolding. In this study, TAT-OP1 was shown to be a useful strategy to efficiently drive denatured OP1 into mouse MC3T3E1 pre-osteoblasts. The correct in vitro protein refolding was verified by the activation of the BMP cascade, while the osteogenic potential of OP1 was demonstrated by increased expression of alkaline phosphatase, osteonectin and osteocalcin.
Product Description
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%