MilliporeSigma
  • Multiscale patterning of a biomimetic scaffold integrated with composite microspheres.
Multiscale patterning of a biomimetic scaffold integrated with composite microspheres.
Small (Weinheim an der Bergstrasse, Germany) (2014-05-29)
Silvia Minardi, Monica Sandri, Jonathan O Martinez, Iman K Yazdi, Xeuwu Liu, Mauro Ferrari, Bradley K Weiner, Anna Tampieri, Ennio Tasciotti
ABSTRACT
The ideal scaffold for regenerative medicine should concurrently mimic the structure of the original tissue from the nano- up to the macroscale and recapitulate the biochemical composition of the extracellular matrix (ECM) in space and time. In this study, a multiscale approach is followed to selectively integrate different types of nanostructured composite microspheres loaded with reporter proteins, in a multi-compartment collagen scaffold. Through the preservation of the structural cues of the functionalized collagen scaffold at the nano- and microscale, its macroscopic features (pore size, porosity, and swelling) are not altered. Additionally, the spatial confinement of the microspheres allows the release of the reporter proteins in each of the layers of the scaffold. Finally, the staged and zero-order release kinetics enables the temporal biochemical patterning of the scaffold. The versatile manufacturing of each component of the scaffold results in the ability to customize it to better mimic the architecture and composition of the tissues and biological systems.
Product Description
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane, Selectophore, ≥99.5%
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Supelco
Rhodamine B, analytical standard
Supelco
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
Sigma-Aldrich
Rhodamine B, suitable for fluorescence