MilliporeSigma
  • Podocalyxin promotes glioblastoma multiforme cell invasion and proliferation via β-catenin signaling.
Podocalyxin promotes glioblastoma multiforme cell invasion and proliferation via β-catenin signaling.
PloS one (2014-10-29)
Yu Liu, Liang Yang, Bo Liu, Yu-Gang Jiang
ABSTRACT
Both podocalyxin (PODX) and β-catenin (β-cat) signaling reportedly play important roles in glioblastoma multiforme (GBM) progression. In this study, we for the first time explored crosstalk between PODX and β-cat signaling in GBM cells, and assessed its impact on GBM cell invasion and proliferation. Stable overexpression of PODX in LN-229 and U-118 MG human GBM cells increased the soluble/intracellular β-cat level, TOPflash luciferase reporter activity, the mRNA levels of β-cat signaling target genes, matrix metalloproteinase 9 (MMP9) expression/activity, and cell invasion and proliferation, which was abolished by selective p38 mitogen-activated protein kinase (MAPK) inhibitor PD169316 and selective β-cat signaling inhibitor CCT031374. On the other hand, stable knockdown of PODX in LN-229 and U-118 MG cells decreased the soluble β-cat level, TOPflash luciferase reporter activity, the mRNA levels of β-cat signaling target genes, MMP9 expression/activity, and cell invasion and proliferation, which was completely reversed by overexpression of a constitutively active β-cat mutant. In addition, overexpression of PODX induced p38 MAPK activity and inactivating phosphorylation of glycogen synthase kinase-3β (GSK-3β) at serine 389 in LN-229 and U-118 MG cells, which was abolished by PD169316, but not CCT031374; knockdown of PODX decreased p38 MAPK activity and inactivating phosphorylation of GSK-3β at serine 389 in both cell lines, which was not significantly affected by overexpression of constitutively active β-cat. In conclusion, this study indicates that PODX promotes GBM cell invasion and proliferation by elevating the soluble β-cat level/β-cat signaling through the p38 MAPK/GSK-3β pathway. Uncovering the PODX/β-cat signaling axis adds new insights not only into the biological functions of PODX and β-cat, but also into the molecular mechanisms underlying GBM progression.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material