MilliporeSigma
  • Optimal attenuation of experimental autoimmune encephalomyelitis by intravenous immunoglobulin requires an intact interleukin-11 receptor.
Optimal attenuation of experimental autoimmune encephalomyelitis by intravenous immunoglobulin requires an intact interleukin-11 receptor.
PloS one (2014-08-01)
Carlyn A Figueiredo, Paulina C Drohomyrecky, Stephen D S McCarthy, Danila Leontyev, Xue-Zhong Ma, Donald R Branch, Shannon E Dunn
ABSTRACT
Intravenous immunoglobulin (IVIg) has been used to treat a variety of autoimmune disorders including multiple sclerosis (MS); however its mechanism of action remains elusive. Recent work has shown that interleukin-11 (IL-11) mRNAs are upregulated by IVIg in MS patient T cells. Both IVIg and IL-11 have been shown to ameliorate experimental autoimmune encephalomyelitis (EAE), an animal model of MS. The objective of this study was to determine whether the protective effects of IVIg in EAE occur through an IL-11 and IL-11 receptor (IL-11R)-dependent mechanism. We measured IL-11 in the circulation of mice and IL-11 mRNA expression in various organs after IVIg treatment. We then followed with EAE studies to test the efficacy of IVIg in wild-type (WT) mice and in mice deficient for the IL-11 receptor (IL-11Rα-/-). Furthermore, we evaluated myelin-specific Th1 and Th17 responses and assessed spinal cord inflammation and demyelination in WT and IL-11Rα-/- mice, with and without IVIg treatment. We also examined the direct effects of mouse recombinant IL-11 on the production of IL-17 by lymph node mononuclear cells. IVIg treatment induced a dramatic surge (>1000-fold increase) in the levels of IL-11 in the circulation and a prominent increase of IL-11 mRNA expression in the liver. Furthermore, we found that IL-11Rα-/- mice, unlike WT mice, although initially protected, were resistant to full protection by IVIg during EAE and developed disease with a similar incidence and severity as control-treated IL-11Rα-/- mice, despite initially showing protection. We observed that Th17 cytokine production by myelin-reactive T cells in the draining lymph nodes was unaffected by IVIg in IL-11Rα-/- mice, yet was downregulated in WT mice. Finally, IL-11 was shown to directly inhibit IL-17 production of lymph node cells in culture. These results implicate IL-11 as an important immune effector of IVIg in the prevention of Th17-mediated autoimmune inflammation during EAE.
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, SAJ special grade, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland