MilliporeSigma
  • Molecular characterization of a thermophilic endo-polygalacturonase from Thielavia arenaria XZ7 with high catalytic efficiency and application potential in the food and feed industries.
Molecular characterization of a thermophilic endo-polygalacturonase from Thielavia arenaria XZ7 with high catalytic efficiency and application potential in the food and feed industries.
Journal of agricultural and food chemistry (2014-12-11)
Tao Tu, Kun Meng, Huoqing Huang, Huiying Luo, Yingguo Bai, Rui Ma, Xiaoyun Su, Pengjun Shi, Peilong Yang, Yaru Wang, Bin Yao
ABSTRACT
Thermophilic endo-polygalacturonases with high catalytic efficiency are of great interest in the food and feed industries. This study identified an endo-polygalacturonase gene (pg7fn) of glycoside hydrolase family 28 in the thermophilic fungus Thielavia arenaria XZ7. Recombinant PG7fn produced in Pichia pastoris is distinguished from other enzyme counterparts by its high functional temperature (60 °C) and specific activity (34382 ± 351 U/mg toward polygalacturonic acid). The enzyme exhibited good pH stability (pH 3.0-8.0) and resistance to pepsin and trypsin digestion and had a significant effect on disaggregation of soybean meal. Addition of 1 U/g PG7fn increased the pectin bioavailability by 19.33%. The excellent properties described above make PG7fn valuable for applications in the food and feed industries. Furthermore, a comparative study showed that N-glycosylation improved the thermostability and catalytic efficiency of PG7fn.
Sigma-Aldrich
Trigalacturonic acid, ≥90% (HPLC)
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard