MilliporeSigma
  • Effect of the sp(2)/sp(3) Ratio in a Hybrid Nanocarbon Thin Film Electrode for Anodic Stripping Voltammetry Fabricated by Unbalanced Magnetron Sputtering Equipment.
Effect of the sp(2)/sp(3) Ratio in a Hybrid Nanocarbon Thin Film Electrode for Anodic Stripping Voltammetry Fabricated by Unbalanced Magnetron Sputtering Equipment.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-07-15)
Hiroyuki Yanagisawa, Ryoji Kurita, Tomoyuki Kamata, Kyoko Yoshioka, Dai Kato, Ayumi Iwasawa, Tetsuya Nakazato, Masaki Torimura, Osamu Niwa
ABSTRACT
The effect of the sp(2)/sp(3) ratio in an unbalanced magnetron sputtered nanocarbon film electrode was studied for determining Cd(2+) and Pb(2+) by anodic stripping voltammetry (ASV). The signal-to-noise ratio in the ASV measurement improved as the sp(3) concentration in the carbon film increased because the noise current decreased with the increasing sp(3) concentration. The detection limits with a carbon film containing 50% sp(3) were 0.25 and 1.0 μg L(-1) for Cd(2+) and Pb(2+) with high repeatability (Cd: 4.6% and Pb: 6.4%, n = 3). For a real sample measurement, a pretreatment system combining a photooxidation reactor and a cation exchange column was used to eliminate the interference from EDTA and Cu(2+), which forms a stable complex or alloy with Cd(2+) and Pb(2+). More than 99% of the interference was eliminated, and accurate signal currents for Cd(2+) and Pb(2+) were successfully obtained with the pretreatment system.
Sigma-Aldrich
Potassium chloride solution, 0.01 M
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, Molecular Biology, ≥99%
Sigma-Aldrich
Hexaammineruthenium(III) chloride, 98%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, Molecular Biology