MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Pharmacokinetics of clomipramine during pregnancy.
European journal of clinical pharmacology (2015-09-30)
P G J Ter Horst, J H Proost, J P Smit, M T Vries, L T W de Jong-van de Berg, B Wilffert
ABSTRACT
Clomipramine is one of the drugs for depression during pregnancy; however, pharmacokinetic data of clomipramine and its active metabolite desmethylclomipramine in this vulnerable period are lacking. In this study, we describe clomipramine and desmethylclomipramine concentrations including their ratios during pregnancy. Second, we describe Center for Epidemiologic Studies Depression scale (CES-D) scores during pregnancy. During 13 pregnancies, every trimester and 3 months after pregnancy, the clomipramine and desmethylclomipramine concentrations were measured with LC-MSMS and the severity of depression was assessed by taking the CES-D score. All concentrations used in our calculations were in fact the ratio between actual plasma concentration (μg/l) and the actual dose (mg). We compared differences in ratios between trimesters by using the Friedman test. Studying 12 women and 13 pregnancies, we found no changes in mean clomipramine concentrations, a statistically significant decrease in mean desmethylclomipramine concentrations (p = 0.014) and a significant decrease in the ratio of desmethylclomipramine/clomipramine mean concentrations during pregnancy (p = 0.014) compared to the post-partum period. Sub-therapeutic concentrations of clomipramine and desmethylclomipramine were found in three patients during whole pregnancy. The mean concentrations of the pharmacologically active metabolite of clomipramine and desmethylclomipramine changes during pregnancy, where a decrease in mean concentrations was found during pregnancy. In case of recurrent disease, we recommend to control clomipramine and its metabolite concentrations, while both are active.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC