MilliporeSigma
  • High gastrointestinal permeability and local metabolism of naringenin: influence of antibiotic treatment on absorption and metabolism.
High gastrointestinal permeability and local metabolism of naringenin: influence of antibiotic treatment on absorption and metabolism.
The British journal of nutrition (2015-06-18)
Naiara Orrego-Lagarón, Miriam Martínez-Huélamo, Anna Vallverdú-Queralt, Rosa M Lamuela-Raventos, Elvira Escribano-Ferrer
ABSTRACT
The present study aims to determine the permeability of naringenin in the stomach, small intestine and colon, to evaluate intestinal and hepatic first-pass metabolism, and to study the influence of the microbiota on the absorption and disposition of naringenin (3.5 μg/ml). A single-pass intestinal perfusion model in mice (n 4-6) was used. Perfusate (every 10 min), blood (at 60 min) and bile samples were taken and analysed to evaluate the presence of naringenin and its metabolites by an HPLC-MS/MS method. To study the influence of the microbiota on the bioavailability of naringenin, a group of animals received the antibiotic rifaximin (50 mg/kg per d) for 5 d, and naringenin permeability was determined in the colon. Naringenin was absorbed well throughout the gastrointestinal tract but mainly in the small intestine and colon (mean permeability coefficient 7.80 (SD 1.54) × 10(-4) cm/s and 5.49 (SD 1.86) × 10(-4) cm/s, respectively), at a level similar to the highly permeable compound, naproxen (6.39 (SD 1.23) × 10(-4) cm/s). According to the high amounts of metabolites found in the perfusate compared to the bile and plasma, naringenin underwent extensive intestinal first-pass metabolism, and the main metabolites excreted were sulfates (84.00 (SD 12.14)%), followed by glucuronides (8.40 (SD 5.67)%). Phase II metabolites were found in all perfusates from 5 min of sampling. Mice treated with rifaximin showed a decrease in naringenin permeability and in the amounts of 4-hydroxyhippuric acid and hippuric acid in the lumen. Naringenin was well absorbed throughout the gastrointestinal tract and its poor bioavailability was due mainly to high intestinal metabolism.
Sigma-Aldrich
L-Methionine, SAJ special grade, ≥98.5%
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
SAFC
L-Methionine
Sigma-Aldrich
Phloroglucinol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG