MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Self-coated interfacial layer at organic/inorganic phase for temporally controlling dual-drug delivery from electrospun fibers.
Self-coated interfacial layer at organic/inorganic phase for temporally controlling dual-drug delivery from electrospun fibers.
Colloids and surfaces. B, Biointerfaces (2015-04-17)
Xin Zhao, Jingwen Zhao, Zhi Yuan William Lin, Guoqing Pan, Yueqi Zhu, Yingsheng Cheng, Wenguo Cui
ABSTRACT
Implantable tissue engineering scaffolds with temporally programmable multi-drug release are recognized as promising tools to improve therapeutic effects. A good example would be one that exhibits initial anti-inflammatory and long-term anti-tumor activities after tumor resection. In this study, a new strategy for self-coated interfacial layer on drug-loaded mesoporous silica nanoparticles (MSNs) based on mussel-mimetic catecholamine polymer (polydopamine, PDA) layer was developed between inorganic and organic matrix for controlling drug release. When the interface PDA coated MSNs were encapsulated in electrospun poly(L-lactide) (PLLA) fibers, the release rates of drugs located inside/outside the interfacial layer could be finely controlled, with short-term release of anti-inflammation ibuprofen (IBU) for 30 days in absence of interfacial interactions and sustained long-term release of doxorubicin (DOX) for 90 days in presence of interfacial interactions to inhibit potential tumor recurrence. The DOX@MSN-PDA/IBU/PLLA hybrid fibrous scaffolds were further found to inhibit proliferation of inflammatory macrophages and cancerous HeLa cells, while supporting the normal stromal fibroblast adhesion and proliferation at different release stages. These results have suggested that the interfacial obstruction layer at the organic/inorganic phase was able to control the release of drugs inside (slow)/outside (rapid) the interfacial layer in a programmable manner. We believe such interface polymer strategy will find applications in where temporally controlled multi-drug delivery is needed.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%