MilliporeSigma
  • Sequential phosphorylation analysis using dye-tethered peptides and microfluidic isoelectric focusing electrophoresis.
Sequential phosphorylation analysis using dye-tethered peptides and microfluidic isoelectric focusing electrophoresis.
Biosensors & bioelectronics (2015-06-09)
Hoseok Choi, Nakchul Choi, Butaek Lim, Tae-Wuk Kim, Simon Song, Young-Pil Kim
ABSTRACT
We report a simple method for analyzing sequential phosphorylation by protein kinases using fluorescent peptide substrates and microfluidic isoelectric focusing (μIEF) electrophoresis. When a dye-labeled peptide substrate was sequentially phosphorylated by two consecutive protein kinases (mitogen-activated protein kinase (MAPK) and glycogen synthase kinase 3 (GSK3)), its differently phosphorylated forms were easily separated and visualized by fluorescent focusing zones in the μIEF channel based on a change in the isoelectric point (pI) by phosphorylation. As a result, ratiometric and quantitative analysis of the fluorescent focusing regions shifted by phosphorylation enabled the analysis of phosphorylation efficiency and the relevant inhibition of protein kinases (MAPK and GSK3) with high simplicity and selectivity. Furthermore, the GSK3 activity in the cell lysates was elucidated by μIEF electrophoresis in combination with immunoprecipitation. Our results suggest that this method has great potential for analyzing the sequential phosphorylation of multiple protein kinases that are implicated in cellular signaling pathways.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of 30 mg
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) disodium salt hydrate, vial of ~1 mg ATP
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade II, ≥97% (HPLC), crystalline, from microbial
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis