MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Characterization of endogenously circulating IGFBP-4 fragments-Novel biomarkers for cardiac risk assessment.
Characterization of endogenously circulating IGFBP-4 fragments-Novel biomarkers for cardiac risk assessment.
Clinical biochemistry (2015-05-31)
Alexey A Konev, Tatyana I Smolyanova, Alexey V Kharitonov, Daria V Serebryanaya, Stanislav V Kozlovsky, Andrey N Kara, Evgeniya E Feygina, Alexey G Katrukha, Alexander B Postnikov
ABSTRACT
Recent findings show that circulating N- and C-terminal fragments of IGF-binding protein-4 (NT-IGFBP-4 and CT-IGFBP-4) can be utilized as biomarkers for cardiac risk assessment in acute coronary syndrome (ACS) patients. The fragments are thought to be the products of pregnancy-associated plasma protein A (PAPP-A)-dependent proteolysis. Two immunoassays for the measurement of IGFBP-4 fragments have been proposed. However, properties of the endogenous IGFBP-4 fragments that could influence the performance of the immunoassays were still not investigated. NT- and CT-IGFBP-4 were extracted from pooled ACS plasma using affinity purification, and their concentrations were measured using sandwich immunoassays utilizing antibodies specific to their proteolytic neo-epitopes or internal epitopes. The extracted fragments were characterized by Western blots (WB) and mass-spectrometry. ACS plasma samples were analyzed by size exclusion chromatography (SEC). Immunoassays utilizing the neo-epitope-specific and the internal epitope-specific antibodies measured equal concentrations of the analyte in the endogenous IGFBP-4 fragments preparations. Only the 18 kDa NT-IGFBP-4 and 14 kDa CT-IGFBP-4 were detected in the WB analysis. Using mass-spectrometry, peaks corresponding to intact non-truncated and non-modified NT-IGFBP-4 (14626 Da) and CT-IGFBP-4 (11346 Da) were observed. The absence of complexed forms of IGFBP-4 in patients' plasma was demonstrated using SEC. Endogenous NT- and CT-IGFBP-4 from ACS patients' plasma correspond to the PAPP-A-derived IGFBP-4 fragments and do not undergo any truncation, modification, or complex formation in the patients' blood. Because of the demonstrated intact state of the circulating IGFBP-4 fragments, the neo-epitope-specific immunoassays perform reliably, allowing further clinical validation of these novel biomarkers.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
2,5-Dihydroxybenzoic acid, suitable for matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%