MilliporeSigma
Expression of immune checkpoint molecules in endometrial carcinoma.
Experimental and therapeutic medicine (2015-12-08)
Jia Liu, Yuling Liu, Wuliang Wang, Chenyang Wang, Yanhong Che
ABSTRACT
The main obstacle in the development of an effective tumor vaccine is the inherent ability of tumors to evade immune responses. Tumors often use common immune mechanisms and regulators to evade the immune system. The present study aimed to analyze the expression levels of indoleamine 2,3-dioxygenase (IDO), programmed death-ligand (PD-L) 1, PD-L2, B7-H4, galectin-1 and galectin-3 in tissue samples from patients with endometrial carcinoma, in order to detect the immunosuppressive environment of endometrial carcinomas. The levels of IDO, PD-L1, PD-L2 and B7-H4 were analyzed by immunohistochemical methods, and the levels of galectin-1 and galectin-3 in tumor lysates were determined using ELISA. PD-L2 was expressed at low levels in the majority of tumor samples. IDO expression was detected in 38, 63 and 43% of primary endometrial carcinoma, recurrent endometrial carcinoma, and metastatic endometrial carcinoma specimens, respectively. Positive expression rates for PD-L1 were 83% in primary endometrial carcinoma, 68% in recurrent endometrial carcinoma, and 100% in metastatic endometrial carcinoma, whereas B7-H4 expression was detected in 100% of both primary endometrial carcinoma and recurrent endometrial carcinoma samples, and in 96% of metastatic endometrial carcinoma specimens. The expression levels of galectin-1 and galectin-3 were not significantly different between the normal and tumor specimens. The results of the present study suggest that the interaction between PD-1/PD-L1 and B7-H4 may be a potential target for immune intervention in the treatment of endometrial carcinoma. Furthermore, the results may provide the basis for immunosuppressant therapy in the treatment of patients with uterine cancer.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)