MilliporeSigma
  • Zein-Based Nanoparticles Improve the Oral Bioavailability of Resveratrol and Its Anti-inflammatory Effects in a Mouse Model of Endotoxic Shock.
Zein-Based Nanoparticles Improve the Oral Bioavailability of Resveratrol and Its Anti-inflammatory Effects in a Mouse Model of Endotoxic Shock.
Journal of agricultural and food chemistry (2015-06-02)
Rebeca Penalva, Irene Esparza, Eneko Larraneta, Carlos J González-Navarro, Carlos Gamazo, Juan M Irache
ABSTRACT
Resveratrol offers pleiotropic health benefits including a reported ability to inhibit lipopolysaccharide (LPS)-induced cytokine production. The aim of this work was to prepare, characterize, and evaluate a resveratrol nanoparticulate formulation based on zein. For this purpose, the oral bioavailability of the encapsulated polyphenol as well as its anti-inflammatory effects in a mouse model of endotoxic shock was studied. The resveratrol-loaded nanoparticles displayed a mean size of 307 ± 3 nm, with a negative zeta potential (-51.1 ± 1.55 mV), and a polyphenol loading of 80.2 ± 3.26 μg/mg. In vitro, the release of resveratrol from the nanoparticles was found to be pH independent and adjusted well to the Peppas-Sahlin kinetic model, suggesting a mechanism based on the combination of diffusion and erosion of the nanoparticle matrix. Pharmacokinetic studies demonstrated that zein-based nanoparticles provided high and prolonged plasma levels of the polyphenol for at least 48 h. The oral bioavailability of resveratrol when administered in these nanoparticles increased up to 50% (19.2-fold higher than for the control solution of the polyphenol). Furthermore, nanoparticles administered daily for 7 days at 15 mg/kg were able to diminish the endotoxic symptoms induced in mice by the intraperitoneal administration of LPS (i.e., hypothermia, piloerection, and stillness). In addition, serum tumor necrosis factor-alpha (TNF-α) levels were slightly lower (approximately 15%) than those observed in the control.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Pricing and availability is not currently available.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 1000
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for residue analysis, JIS 300
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for residue analysis, JIS 5000
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, suitable for chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Pricing and availability is not currently available.
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Pricing and availability is not currently available.