MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • AAA-ATPase NVL2 acts on MTR4-exosome complex to dissociate the nucleolar protein WDR74.
AAA-ATPase NVL2 acts on MTR4-exosome complex to dissociate the nucleolar protein WDR74.
Biochemical and biophysical research communications (2015-10-13)
Nobuhiro Hiraishi, Yo-ichi Ishida, Masami Nagahama
ABSTRACT
Nuclear VCP-like 2 (NVL2) is a chaperone-like nucleolar ATPase of the AAA (ATPase associated with diverse cellular activities) family, which exhibits a high level of amino acid sequence similarity with the cytosolic AAA-ATPase VCP/p97. These proteins generally act on macromolecular complexes to stimulate energy-dependent release of their constituents. We previously showed that NVL2 interacts with RNA processing/degradation machinery containing an RNA helicase MTR4/DOB1 and an exonuclease complex, nuclear exosome, and involved in the biogenesis of 60S ribosomal subunits. These observations implicate NVL2 as a remodeling factor for the MTR4-exosome complex during the maturation of pre-ribosomal particles. Here, we used a proteomic screen and identified a WD repeat-containing protein 74 (WDR74) as a factor that specifically dissociates from this complex depending on the ATPase activity of NVL2. WDR74 shows weak amino acid sequence similarity with the yeast ribosome biogenesis protein Nsa1 and is co-localized with NVL2 in the nucleolus. Knockdown of WDR74 decreases 60S ribosome levels. Taken together, our results suggest that WDR74 is a novel regulatory protein of the MTR4-exsosome complex whose interaction is regulated by NVL2 and is involved in ribosome biogenesis.
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis