MilliporeSigma
  • MtrA, an essential response regulator of the MtrAB two-component system, regulates the transcription of resuscitation-promoting factor B of Mycobacterium tuberculosis.
MtrA, an essential response regulator of the MtrAB two-component system, regulates the transcription of resuscitation-promoting factor B of Mycobacterium tuberculosis.
Microbiology (Reading, England) (2015-04-03)
Arun Kumar Sharma, Ayan Chatterjee, Shamba Gupta, Rajdeep Banerjee, Sukhendu Mandal, Jayanta Mukhopadhyay, Joyoti Basu, Manikuntala Kundu
ABSTRACT
The resuscitation-promoting factors of Mycobacterium tuberculosis are hydrolytic enzymes, which are required for resuscitation of dormant cells. RpfB, a peptidoglycan remodelling enzyme similar to the lytic transglycosylase of Escherichia coli, is required for reactivation of M. tuberculosis from chronic infection in vivo, underscoring the need to understand its transcriptional regulation. Here, we identified the transcriptional and translational start points of rpfB, and suggested from rpf promoter-driven GFP expression and in vitro transcription assays that its transcription possibly occurs in a SigB-dependent manner. We further demonstrated that rpfB transcription is regulated by MtrA - the response regulator of the essential two-component system MtrAB. Association of MtrA with the rpfB promoter region in vivo was confirmed by chromatin immunoprecipitation analysis. Electrophoretic mobility shift assays (EMSAs) revealed a loose direct repeat sequence associated with MtrA binding. Binding of MtrA was enhanced upon phosphorylation. MtrA could be pulled down from lysates of M. tuberculosis using a biotinylated DNA fragment encompassing the MtrA-binding site on the rpfB promoter, confirming that MtrA binds to the rpfB promoter. Enhanced GFP fluorescence driven by the rpfB promoter, upon deletion of the MtrA-binding site, and repression of rpfB expression, upon overexpression of MtrA, suggested that MtrA functions as a repressor of rpfB transcription. This was corroborated by EMSAs showing diminished association of RNA polymerase (RNAP) with the rpfB promoter in the presence of MtrA. In vitro transcription assays confirmed that MtrA inhibits RNAP-driven rpfB transcription.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C