MilliporeSigma
  • In vitro inhibitory effects of dihydromyricetin on human liver cytochrome P450 enzymes.
In vitro inhibitory effects of dihydromyricetin on human liver cytochrome P450 enzymes.
Pharmaceutical biology (2017-06-16)
Lu Liu, Sen Sun, Hongbing Rui, Xiaohua Li
ABSTRACT
Dihydromyricetin (DHM) is the most abundant and active flavonoid component isolated from Ampelopsis grossedentata (Hand-Mazz) W.T. Wang (Vitaceae) and it possesses numerous pharmacological activities. However, whether DHM affects the activity of human liver cytochrome P450 (CYP) enzymes remains unclear. The inhibitory effects of DHM on eight human liver CYP isoforms (i.e., 1A2, 3A4, 2A6, 2E1, 2D6, 2C9, 2C19 and 2C8) were investigated in vitro using human liver microsomes (HLMs). The results showed that DHM could inhibit the activity of CYP3A4, CYP2E1 and CYP2D6, with IC The in vitro studies of DHM with CYP isoforms indicate that DHM has the potential to cause pharmacokinetic drug interactions with other co-administered drugs metabolized by CYP3A4, CYP2E1 and CYP2D6. Further clinical studies are needed to evaluate the significance of this interaction.
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Dextrorphan
Pricing and availability is not currently available.
Sigma-Aldrich
4-Hydroxymephenytoin, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Pricing and availability is not currently available.