Skip to Content
MilliporeSigma

8.43978

Potassium-tert-butylate

(1M solution in tert-butanol) for synthesis

Synonym(s):

Potassium tert-butoxide

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352202

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Potassium-tert-butylate, (1M solution in tert-butanol) for synthesis

form

liquid

transition temp

flash point 14 °C (Information refers to the main component.)

density

0.820 g/cm3

storage temp.

2-30°C

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8.049188.18374659878
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

powder

form

powder

form

solid

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

density

0.820 g/cm3

density

1.19 g/cm3 at 20 °C

density

-

density

-

transition temp

flash point 14 °C (Information refers to the main component.)

transition temp

-

transition temp

-

transition temp

-

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 2 - Skin Corr. 1 - Water-react 1

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

WGK 1

flash_point_f

57.2 °F - Information refers to the main ingredient.

flash_point_c

14 °C - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service