Select a Size
$242.00
$435.00
$242.00
About This Item
Skip To
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AAAGGTCCATCCAAGCTGAATGATCGCGCTGACTCAAGAAGAAGCCTTTGGGACCAAGGAAACTTTCCCCTGATCATCAAGAATCTTAAGATAGAAGACTCAGATACTTACATCTGTGAAGTGGAGGACCAGAAGGAGGAGGTGCAATTGCTAGTGTTCGGATTGACTGCCAACTCTGACACCCACCTGCTTCAGGGGCAGAGCCTGACCCTGACCTTGGAGAGCCCCCCTGGTAGTAGCCCCTCAGTGCAATGTAGGAGTCCAAGGGGTAAAAACATACAGGGGGGGAAGACCCTCTCCGTGTCTCAGCTGGAGCTCCAGGATAGTGGCACCTGGACATGCACTGTCTTGCAGAACCAGAAGAAGGTGGAGTTCAAAATAGACATCGTGGTGCTAGCTTTCCAGAAGGCCTCCAGCATAGTCTATAAGAAAGAGGGGGAACAGGTGGAGTTCTCCTTCCCACTCGCCTTTACAG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
1 of 4
This Item | EHU148081 | EHU050931 | EHU041301 |
|---|---|---|---|
| Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. | Ensembl | human accession no. |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information human ... CD4(920), CD4(920) | Gene Information human ... CD8B(926), CD8B(926) | Gene Information human ... CD14(929), CD14(929) | Gene Information human ... CD9(928), CD9(928) |
| esiRNA cDNA target sequence AAAGGTCCATCCAAGCTGAATGATCGCGCTGACTCAAGAAGAAGCCTTTGGGACCAAGGAAACTTTCCCCTGATCATCAAGAATCTTAAGATAGAAGACTCAGATACTTACATCTGTGAAGTGGAGGACCAGAAGGAGGAGGTGCAATTGCTAGTGTTCGGATTGACTGCCAACTCTGACACCCACCTGCTTCAGGGGCAGAGCCTGACCCTGACCTTGGAGAGCCCCCCTGGTAGTAGCCCCTCAGTGCAATGTAGGAGTCCAAGGGGTAAAAACATACAGGGGGGGAAGACCCTCTCCGTGTCTCAGCTGGAGCTCCAGGATAGTGGCACCTGGACATGCACTGTCTTGCAGAACCAGAAGAAGGTGGAGTTCAAAATAGACATCGTGGTGCTAGCTTTCCAGAAGGCCTCCAGCATAGTCTATAAGAAAGAGGGGGAACAGGTGGAGTTCTCCTTCCCACTCGCCTTTACAG | esiRNA cDNA target sequence ATGGCAACTCAGTCCTCCAGCAGACCCCTGCATACATAAAGGTGCAAACCAACAAGATGGTGATGCTGTCCTGCGAGGCTAAAATCTCCCTCAGTAACATGCGCATCTACTGGCTGAGACAGCGCCAGGCACCGAGCAGTGACAGTCACCACGAGTTCCTGGCCCTCTGGGATTCCGCAAAAGGGACTATCCACGGTGAAGAGGTGGAACAGGAGAAGATAGCTGTGTTTCGGGATGCAAGCCGGTTCATTCTCAATCTCACAAGCGTGAAGCCGGAAGACAGTGGCATCTACTTCTGCATGATCGTCGGGAGCCCCGAGCTGACCTTCGGGAAGGGAACTCAGCTGAGTGTGGTTGATTTCCTTCCCACCACTGCCCAGCCCACCAAGAAGTCCACCCTCAAG | esiRNA cDNA target sequence GCCGCTGTGTAGGAAAGAAGCTAAAGCACTTCCAGAGCCTGTCCGGAGCTCAGAGGTTCGGAAGACTTATCGACCATGGAGCGCGCGTCCTGCTTGTTGCTGCTGCTGCTGCCGCTGGTGCACGTCTCTGCGACCACGCCAGAACCTTGTGAGCTGGACGATGAAGATTTCCGCTGCGTCTGCAACTTCTCCGAACCTCAGCCCGACTGGTCCGAAGCCTTCCAGTGTGTGTCTGCAGTAGAGGTGGAGATCCATGCCGGCGGTCTCAACCTAGAGCCGTTTCTAAAGCGCGTCGATGCGGACGCCGACCCGCGGCAGTATGCTGACACGGTCAAGGCTCTCCGCGTGCGGCGGCTCACAGTGGGAGCCGCACAGGTTCCTGCTCAGCTACTGGTAGGCGCCCTGCGTGTGCTAGCGTACTCCCGCCTCAAGGAACTGACGCTCGAGGACCTA | esiRNA cDNA target sequence TTGGACTATGGCTCCGATTCGACTCTCAGACCAAGAGCATCTTCGAGCAAGAAACTAATAATAATAATTCCAGCTTCTACACAGGAGTCTATATTCTGATCGGAGCCGGCGCCCTCATGATGCTGGTGGGCTTCCTGGGCTGCTGCGGGGCTGTGCAGGAGTCCCAGTGCATGCTGGGACTGTTCTTCGGCTTCCTCTTGGTGATATTCGCCATTGAAATAGCTGCGGCCATCTGGGGATATTCCCACAAGGATGAGGTGATTAAGGAAGTCCAGGAGTTTTACAAGGACACCTACAACAAGCTGAAAACCAAGGATGAGCCCCAGCGGGAAACGCTGAAAGCCATCCACTATGCGTTGAACTGCTGTGGTTTGGCTGGGGGCGTGGAACAGTTTATCTCAGACATCTGCCCCAAGAAGGACGTACTCGAAACCTTCACCGTGAAGTCCTGTCCTGATGCCATC |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service