Skip to Content
MilliporeSigma

EHU151881

MISSION® esiRNA

targeting human SCN5A

Sign In to View Organizational & Contract Pricing

Select a Size

20 μG

$242.00

50 μG

$435.00

$242.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAACGCCTTGTATGTCCTCAGTCCCTTCCACCCCATCCGGAGAGCGGCTGTGAAGATTCTGCTTTCCTTGACCCCACGCACGCTCTTCAACATGCTCATCATGTGCACCATCCTCACCAACTGCGTGTTCATGGCCCAGCACGACCCTCCACCCTGGACCAAGTATGTCGAGTACACCTTCACCGCCATTTACACCTTTGAGTCTCTGGTCAAGATTCTGGCTCGAGGCTTCTGCCTGCACGCGTTCACTTTCCTTCGGGACCCATGGAACTGGCTGGACTTTAGTGTGATTATCATGGCATACACAACTGAATTTGTGGACCTGGGCAATGTCTCAGCCTTACGCACCTTCCGAGTCCTCCGGGCCCTGAAAACTATATCAGTCATTTCAGGGCTGAAGACCATCGTGGGGGCCCTGATCCAGTCTGTGAAGAAGCTGGCTGATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU113551EHU141361EHU066451
esiRNA cDNA target sequence

CAACGCCTTGTATGTCCTCAGTCCCTTCCACCCCATCCGGAGAGCGGCTGTGAAGATTCTGCTTTCCTTGACCCCACGCACGCTCTTCAACATGCTCATCATGTGCACCATCCTCACCAACTGCGTGTTCATGGCCCAGCACGACCCTCCACCCTGGACCAAGTATGTCGAGTACACCTTCACCGCCATTTACACCTTTGAGTCTCTGGTCAAGATTCTGGCTCGAGGCTTCTGCCTGCACGCGTTCACTTTCCTTCGGGACCCATGGAACTGGCTGGACTTTAGTGTGATTATCATGGCATACACAACTGAATTTGTGGACCTGGGCAATGTCTCAGCCTTACGCACCTTCCGAGTCCTCCGGGCCCTGAAAACTATATCAGTCATTTCAGGGCTGAAGACCATCGTGGGGGCCCTGATCCAGTCTGTGAAGAAGCTGGCTGATG

esiRNA cDNA target sequence

TCAAAAATGGGTACGGCTTCGTGGAGTTCGAGGACTCCCGCGACGCCGACGACGCCGTTTACGAGCTGAACGGCAAGGAGCTCTGCGGCGAGCGCGTGATCGTAGAGCACGCCCGGGGCCCGCGTCGCGATCGCGACGGCTACAGCTACGGAAGCCGCAGTGGTGGAGGTGGATACAGCAGTCGGAGAACATCTGGCAGAGACAAATACGGACCACCTGTTCGTACAGAATACAGGCTTATTGTAGAAAATCTTTCTAGTCGGTGCAGTTGGCAAGATTTAAAGGATTTTATGCGACAAGCAGGTGAAGTAACCTATGCGGATGCCCACAAGGAACGAACAAATGAGGGTGTAATTGAGTTTCGCTCCTACTCTGACATGAAGCGTGCTTTGGACAAACTGGATGGCACAGAAA

esiRNA cDNA target sequence

GAGTCTCAGCCGAAGAGAGCTGGGCTGGGCCCCAGCCCCTTGAGACTCTGGACACCTGAACCAGACCTGCCAGGGCTGTGCGATCTCTTGGCCTGGTCCTTGCAGCTGTGGCAGCAGCAGGCTCCCCAGCGGCCCAGGGTGGGCCAGACCAGCAGCCCAGGAAGCAGCACACGCGGCCGTGGGGAGGCAGGCACCGGGCATGTCGGCGCCTCTGGTCAAACCACCTACACTGCCTGGGGTGGGTCTCAAGGAGGCCCGGGGCGGAGGGGGGTTCCCGCGTGCACACGAGTGCGGCTGGACGTGCCGACACGCGGTGATGTACCCATGCTCCGTGTGTCTGTGTCTGCATGTCCACACGTCTGATGCACCTGTGTACGTGTGTCAAGCCTAGCCACCTCAG

esiRNA cDNA target sequence

CTGCTTCCAAGACCACATGATCCGTAACTGCAACTGTGGCCACTACCTGTACCCACTGCCCCGTGGGGAGAAATACTGCAACAACCGGGACTTCCCAGACTGGGCCCATTGCTACTCAGATCTACAGATGAGCGTGGCGCAGAGAGAGACCTGCATTGGCATGTGCAAGGAGTCCTGCAATGACACCCAGTACAAGATGACCATCTCCATGGCTGACTGGCCTTCTGAGGCCTCCGAGGACTGGATTTTCCACGTCTTGTCTCAGGAGCGGGACCAAAGCACCAATATCACCCTGAGCAGGAAGGGAATTGTCAAGCTCAACATCTACTTCCAAGAATTTAACTATCGCACCATTGAAGAATCAGCAGCCAATAACATCGTCTGGCTGCTCTCGAATCTGGGTGGCCAGTTTGGCTTCTGGATGGGGGGCTCTGTGCTGTGCCTCATCGAGTTTGGGGAGATC

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

human ... SCN5A(6331), SCN5A(6331)

Gene Information

human ... SRSF6(6431), SFRS6(6431)

Gene Information

human ... SCNN1D(6339), SCNN1D(6339)

Gene Information

human ... SCNN1B(6338), SCNN1B(6338)

Ensembl | human accession no.

ENSG00000183873

Ensembl | human accession no.

ENSG00000124193

Ensembl | human accession no.

ENSG00000162572

Ensembl | human accession no.

ENSG00000168447

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Yang et al.
Journal of cellular physiology, 235(4), 3950-3972 (2019-10-16)
Ion channels can regulate the plasma membrane potential (Vm ) and cell migration as a result of altered ion flux. However, the mechanism by which Vm regulates motility remains unclear. Here, we show that the Nav 1.5 sodium channel carries
Wan-Lin Lo et al.
Nature immunology, 13(9), 880-887 (2012-07-31)
The sustained entry of Ca(2+) into CD4(+)CD8(+) double-positive thymocytes is required for positive selection. Here we identified a voltage-gated Na(+) channel (VGSC) that was essential for positive selection of CD4(+) T cells. Pharmacological inhibition of VGSC activity inhibited the sustained
Jie Zhang et al.
Acta biochimica et biophysica Sinica, 51(6), 562-570 (2019-05-30)
The protein voltage-gated sodium channel Nav1.5 is highly upregulated in various types of cancer and, in general, promotes cancer cell invasiveness and metastatic progression. A previous study found that Nav1.5 was highly expressed in poorly differentiated oral squamous cell carcinoma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service