Select a Size
$242.00
$435.00
$242.00
About This Item
Skip To
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TTCAAGATCCAGCACTGTCGCCAGCCTCGATACAGACTCCACTAAAAGCTCAGGACAGAGTAACAGCAATTTAGACACATGTGCTGAGTTCCGAATAAAGTATGTTGGTGCCATCGAGAAACTGGCCGTGTCTGAGGGGAAGAGCCTTGAAGGACCACTAGACCTGATAAATTACATAGATGTCGCCCAGCAAGATGGAAAGTTGCCTTTTGTGCCTTTGGAAGAGGAGTTCATTCTGGGCGTTTCTAAGTACGGCATAAAAGTCTCGACCACGGATCAGCATGGTGTTCTGCATAGGCATGCCCTGTATCTAATCATCCGGATGGTGTGTTACGATGACGGCCTGGGAGCTGGGAAAAGCTTGCTGGCACTCAAGACCACAGATGCAAGCAACGAAGAGTACAGCCTGTGGGTTTACCAGTGTAACAGCCTGGAGCA
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... ITGB1BP1(16413), Itgb1bp1(16413)
Related Categories
1 of 4
This Item | EMU017091 | EMU051281 | EMU050691 |
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information mouse ... ITGB1BP1(16413), Itgb1bp1(16413) | Gene Information mouse ... ITGB5(16419), Itgb5(16419) | Gene Information mouse ... Itgav(16410) | Gene Information mouse ... Itgb3(16416) |
| esiRNA cDNA target sequence TTCAAGATCCAGCACTGTCGCCAGCCTCGATACAGACTCCACTAAAAGCTCAGGACAGAGTAACAGCAATTTAGACACATGTGCTGAGTTCCGAATAAAGTATGTTGGTGCCATCGAGAAACTGGCCGTGTCTGAGGGGAAGAGCCTTGAAGGACCACTAGACCTGATAAATTACATAGATGTCGCCCAGCAAGATGGAAAGTTGCCTTTTGTGCCTTTGGAAGAGGAGTTCATTCTGGGCGTTTCTAAGTACGGCATAAAAGTCTCGACCACGGATCAGCATGGTGTTCTGCATAGGCATGCCCTGTATCTAATCATCCGGATGGTGTGTTACGATGACGGCCTGGGAGCTGGGAAAAGCTTGCTGGCACTCAAGACCACAGATGCAAGCAACGAAGAGTACAGCCTGTGGGTTTACCAGTGTAACAGCCTGGAGCA | esiRNA cDNA target sequence GGTTTCGGGTCTTTTGTTGACAAGGACATCTCTCCTTTCTCCTACACGGCACCGAGATACCAGACCAATCCGTGTATTGGTTACAAGTTATTCCCCAACTGCGTCCCCTCCTTCGGGTTCCGGCATCTGCTGCCTCTCACAGACAGAGTCGACAGCTTCAACGAGGAAGTGAGGAAGCAGAGGGTGTCCCGGAACCGAGATGCCCCCGAGGGGGGGTTTGATGCGGTCCTCCAGGCTGCTGTCTGCAAGGAGAAGATCGGATGGCGAAAAGATGCTCTGCACTTGCTGGTGTTCACAACAGACGATGTGCCCCACATCGCACTGGATGGAAAACTGGGTGGCCTGGTCCAGCCCCACGATGGCCAGTGTCACCTGAATGAAGCCAATGAGTACACAGCCTCTAACCAGATGGACTATCCATCGCTTGCCTTG | esiRNA cDNA target sequence GTCCGACCACCTCAAGAAGAGCAAGAACGAGAACAGCTTCAGCCTCATGAGAATGGCGAAGGAAACTCTGAAACCTGAGGTGCTTCTTAAGTGGCGCCACTCTGTCCGCTCCAAGTTCACCACCGTGCTGGTGATTTCAACATCTCTCCTGAAGAGTGAAATCCCAGGACTGTATTGCTGAGAGCACCCAGCAGTCTCAGTTTATCCACAAAATTGAAATGGATATTTGTCGACTTCTCTTTATGAATAAGTTCAGGTTACATTTACTTCGCCCACACTGACGTGTGTTCTTAGGGACTTAAGTGATGGAATTTCAAGTGATTACCTTTGTTCAGTGTACATAAGACAGACAGCACCGAGTCAGCACGTTACATAGCATAGTACCTCTTCAGTTACTGTTCCTAATTGACTGTGTGGGGTTTCTCTCTTCGGCGTTACGT | esiRNA cDNA target sequence CTGGTAAAAACGCCGTGAATTGTACCTACAAGAACGAGGATGACTGTGTCGTCAGATTCCAGTACTACGAAGACACCAGTGGGAGGGCAGTCCTCTATGTGGTGGAAGAGCCTGAGTGTCCCAAGGGTCCTGATATCCTGGTGGTACTGCTGTCAGTGATGGGGGCCATCCTGCTCATTGGCCTTGCTACTCTGCTCATCTGGAAGCTACTCATCACCATTCATGACCGGAAGGAATTTGCTAAATTTGAGGAAGAACGAGCCAGAGCTAAGTGGGACACAGCAAACAACCCGCTGTATAAAGAGGCCACCTCCACCTTCACCAATATCACCTACCGGGGGACTTAATGAGACCACTTCAGATGACGCCAGGACCGTGGGACTCTCTGCCATTATGTTTACAGAGGACAATTTGTGTGTGTGTGTATGGGTGGGA |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service