Select a Size
$242.00
$435.00
$242.00
About This Item
Skip To
description
Powered by Eupheria Biotech
Quality Level
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GTGTTCCTGGGAGAGCAGAGCGTTGGAAAGACGTCCTTGATCACCCGATTCATGTATGACAGTTTTGACAACACCTATCAGGCAACAATTGGCATTGACTTCTTATCAAAAACAATGTACTTGGAGGATAGAACCGTGCGATTGCAATTATGGGACACAGCAGGTCAAGAGCGGTTCAGGAGCTTGATTCCTAGCTACATTCGAGACTCCACTGTGGCAGTTGTTGTTTATGATATCACAAATGTTAACTCATTCCAGCAAACTACAAAATGGATTGATGATGTCAGAACAGAGAGAGGAAGTGATGTCATCATCATGCTAGTAGGAAATAAAACAGATCTTGCTGATAAGAGGCAAGTGTCAATTGAGGAGGGAGAGAGGAAAGCCAAAGAGCTGAATGTTATGTTTATTGAAACCAGTGCAAAAGCAGGATACAATGTAAAGCAGCTTTTCCGGCGTGTTGCTGCAGCTTTACCTGGAATGGAAAGCACAC
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... RAB6(19346), Rab6(19346)
1 of 4
This Item | EMU169731 | EMU150241 | EMU012411 |
|---|---|---|---|
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. | Ensembl | mouse accession no. |
| esiRNA cDNA target sequence GTGTTCCTGGGAGAGCAGAGCGTTGGAAAGACGTCCTTGATCACCCGATTCATGTATGACAGTTTTGACAACACCTATCAGGCAACAATTGGCATTGACTTCTTATCAAAAACAATGTACTTGGAGGATAGAACCGTGCGATTGCAATTATGGGACACAGCAGGTCAAGAGCGGTTCAGGAGCTTGATTCCTAGCTACATTCGAGACTCCACTGTGGCAGTTGTTGTTTATGATATCACAAATGTTAACTCATTCCAGCAAACTACAAAATGGATTGATGATGTCAGAACAGAGAGAGGAAGTGATGTCATCATCATGCTAGTAGGAAATAAAACAGATCTTGCTGATAAGAGGCAAGTGTCAATTGAGGAGGGAGAGAGGAAAGCCAAAGAGCTGAATGTTATGTTTATTGAAACCAGTGCAAAAGCAGGATACAATGTAAAGCAGCTTTTCCGGCGTGTTGCTGCAGCTTTACCTGGAATGGAAAGCACAC | esiRNA cDNA target sequence GCAGTTTGCACTTTAATTGATGGTAGTTCAGTCAAGATACAGGTTTTTTTGTGTGTGGGGGGGGGGGGGTTGATTGATTTACCTATCCTTCAACCGTTTTAATTAAATGGAATCTACAATGAATGAGGTTTGGCCCTATAGAAGACAGTAGCAGTGGCAGCTACTCACACCCAGTCTCCACTGCTAGCTTCCTGACTGATTCATACCTCTATTCTTGGGTATAGGTTTGTCTGAGCAACTGGGGAGTGGCCTCAGAATAGCATGGGCCATGGTAGTGGAGTAAAGAAAGGTCAGGCAAAAGGAGTGTTCTATCTTCCCAGGGTTCACATTCAGTTTGCTATGTCTTACCATGTCTTTTCTACTTCCCTGTAAATACGTATAAGTCTTTATTCTCTGTACAGTCTGCTCTGCCTCCTGGCTCCCATTAGCCTTCTTTTCTTCTCCTTTTGTTAATATCTTGGGTTTAGTTCCTCCTTCATTCCCCCACTTACTCTTACTTAATCCGTGTACCACTAGGG | esiRNA cDNA target sequence GGGATCCCGAGAACTTCCCTTTTGTTGTGTTGGGAAACAAGATTGACCTGGAAAACAGACAAGTGGCCACAAAGAGGGCACAGGCTTGGTGCTACAGCAAAAACAACATTCCTTACTTCGAGACCAGTGCCAAGGAGGCCATCAATGTGGAGCAGGCCTTCCAGACAATTGCTCGGAATGCCCTTAAACAGGAAACAGAAGTGGAACTGTACAATGAATTCCCTGAACCCATCAAACTGGACAAGAATGACCGGGCCAAGGCCTCCGCAGAAAGCTGCAGTTGTTGAAGGGGCAGTGAGCACAGAGTCCTTCACAGACCAAGAACACACACGTAGGCCTTCAATATGCGTCCCTCCTCCTCCAAACAGACCAGAACGTGAGCTCTCACATCCAGCTGCCAAAAGAAACTCCACCGAACAGTTACCCAACACACACACATACACAAAACACACTCGTGCACACAAAAACACATACACACAACACACACACAGCAAGCAAACTCCAGCCTGT | esiRNA cDNA target sequence AGTGACCTGCTGGAGGAAGACAGGCGGCGCCGTCGTGTAGACTTCCATGATGTTCAGGACTATGCCGATAATATCAAAGCCCAACTCTTTGAAACATCCAGCAAGACAGGCCAAAGTGTGGATGAACTCTTCCAGAAAGTGGCTGAGGATTACGTCAGTGTGGCTGCTTTCCAGGTGATGACAGAGGACAAAGGTGTGGATTTGAGCCAGAAGGCAAACCCTTACTTCTACAGCTGTTGTCATCACTGAGTCACCAGTCATCTGGCCCAGTGGAATTAGATGAATTCCCAGAGGGGCTGGACCTGACTCTTGTCTGGGCTGGAATGGTCAAGCGTCTGAGCTATTCCAGGTGCCTCTCACAGCAGAGGTGGCACCTGCCTGTGCTGGCCCATGGAACGGAGGCAGTATTGGGCTGACTGTGGG |
| Gene Information mouse ... RAB6(19346), Rab6(19346) | Gene Information mouse ... RAB5B(19344), Rab5b(19344) | Gene Information mouse ... RAB7(19349), Rab7(19349) | Gene Information mouse ... RAB24(19336), Rab24(19336) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| NCBI accession no. | NCBI accession no. | NCBI accession no. | NCBI accession no. |
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service