Skip to Content
MilliporeSigma

EP951031704

Eppendorf® Deepwell Plate 384/200 μL

Protein LoBind, non-sterile, volume 200 μL, white border, clear wells, pkg of 120 plates (10 bags x 12 plates)

Synonym(s):

low protein binding multiplate

Sign In to View Organizational & Contract Pricing.

Select a Size

120 EA

$2,620.00

$2,620.00


Check Cart for Availability


About This Item

UNSPSC Code:
41122107
Material:
clear wells
Size:
384 well
Sterility:
non-sterile
Binding type:
Protein low binding
Feature:
Protein LoBind

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Eppendorf® Deepwell Plate 384/200 μL, Protein LoBind, non-sterile, volume 200 μL, white border, clear wells, pkg of 120 plates (10 bags x 12 plates)

material

clear wells

sterility

non-sterile

feature

Protein LoBind

packaging

pkg of 120 plates (10 bags x 12 plates)

manufacturer/tradename

Eppendorf® 951031704

technique(s)

PCR: suitable

size

384 well

volume

200 μL

color

white border

binding type

Protein low binding

Looking for similar products? Visit Product Comparison Guide

Application

Protein analysis and storage

Features and Benefits

  • OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
  • RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • Minimal remaining volume and high uniformity from well to well
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Easy and reliable stacking
  • g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
  • High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
  • Autoclavable (121°C, 20 min)
  • Available with barcode. Additional details available at: www.eppendorf.com/barcode
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Unique OptiTrack® matrix: 30 % faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing in plates

General description

Eppendorf® Deepwell Plates 96 and 384 are high-performance plates for all manual and automated applications – from sample storage at -86 ºC to DNA denaturation at 100 ºC. Manufactured from pure polypropylene (PP), these plates offer innovative features: More dependability. More efficiency. More stability. The plates are available with a white, yellow, green, or blue border. For easy identification of wells, a high-contrast alphanumeric label is laser-engraved in the border. This OptiTrack® matrix allows you to find your sample up to 30 % more quickly while reducing pipetting errors. With all of these sophisticated features, these plates set new standards in your laboratory.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service