MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Bond Strength of Experimental Low-viscosity Resin Materials to Early Enamel Caries Lesions: Effect of Diluent/Solvent Addition.
Bond Strength of Experimental Low-viscosity Resin Materials to Early Enamel Caries Lesions: Effect of Diluent/Solvent Addition.
The journal of adhesive dentistry (2015-04-02)
Tatiany Gabrielle Freire Araújo, Ravana Angelini Sfalcin, Giovana Spagnolo Albamonte de Araújo, Roberta Caroline Brusch Alonso, Regina Maria Puppin-Rontani
ABSTRACT
To evaluate the effect of different concentrations of monomers and solvents/diluents on the microtensile bond strength (μTBS) bond strength of experimental low-viscosity resins (infiltrants) to enamel caries-like lesions (ECLL). Flat enamel blocks obtained from sound human third molars were submitted to ECLL formation and randomly distributed into 9 groups (n = 10): G1: TEG-DMA 100%; G2: TEG-DMA 80%, ethanol 20%; G3; TEG-DMA 80%, HEMA 20%; G4: TEG-DMA 75%, UDMA 25%; G5: TEG-DMA 60%, UDMA 20%, ethanol 20%; G6: TEG-DMA 60%, UDMA 20%, HEMA 20%; G7: TEG-DMA 75%, bis-EMA 25%; G8: TEG-DMA 60%, bis-EMA 20%, ethanol 20%; G9: TEG-DMA 60%, bis-EMA 20%, HEMA 20%. After etching with 37% phosphoric acid for 60 s, experimental infiltrants were actively applied and photocured for 60 s, then stored in 100% humidity (24 h, 37°C). Hourglass-shaped specimens were obtained and the μTBS test performed (MPa). The fracture patterns were assessed by SEM. Data were submitted to two way-ANOVA and Tukey's tests (α = 0.05). The highest μTBS value was observed for G4 (TEG-DMA/UDMA, 19.18 MPa) and the lowest for G5 (TEG-DMA/UDMA/ethanol, 9.00 MPa). A significant decrease in μTBS was observed for all groups containing ethanol (G2, G5, and G8). The addition of HEMA did not affect μTBS values. Most groups showed a high frequency of mixed failure between infiltrant and enamel. The addition of bis-EMA or UDMA to TEG-DMA-based infiltrants did not improve bond strength to carious enamel. The ethanol addition negatively affected the bonding strength of infiltrants to enamel caries-like lesions, regardless of the resin matrix composition of the infiltrant.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Camphorquinone, 97%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 80-120 ppm MEHQ as inhibitor, 95%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Triethylene glycol dimethacrylate, contains 200 ppm monomethyl ether hydroquinone as inhibitor, 99%