Targeted Protein Degradation
Targeted protein degradation (TPD) is a novel drug discovery strategy that eradicates problem proteins from cells by hijacking cellular degradation processes with small molecules. One of these processes is the ubiquitin-proteasome pathway, an intracellular quality control mechanism that utilizes E3 ubiquitin ligases and associated partners to seek out damaged or misfolded proteins for degradation via the proteasome or “cellular trash can.” Proteolysis-targeting chimeras (PROTAC® degraders) are heterobifunctional molecules containing a disease target warhead and E3 ligase ligand tethered together by a linker.
When added to cells, they recruit the target protein to the E3 ligase, hijacking the native degradation pathway to remove the disease target. This protein silencing approach delivers phenotypes that have been called the chemical equivalent to CRISPR – yet with small-molecule control.
Featured Categories
Protease reagents and resources for endo- and exopeptidase cleavage application needs and resources for your protein cleavage workflow.
Duolink® Proximity Ligation Assay (PLA) technology for detecting and quantifying protein-protein interactions.
Pathogen testing is a key component and critical success factor in effective food safety and process control programs.
We have a comprehensive offering of high-purity amino acids and derivatives important to metabolomic pathways...
PROTAC®-mediated degradation of a target provides many advantages to traditional occupancy-based inhibition by small molecules. Degraders developed from leads, warheads, or inhibitors demonstrate improved cellular responses and target selectivity. As the degrader is not eliminated alongside the target, it is recycled, operating catalytically at lower doses. Lastly, where ~80% of protein targets have been deemed intractable to traditional small molecules due to a lack of defined binding cavities or active sites (“undruggables”), lower affinity binders have been successfully developed into successful degraders by their ability to recruit diverse targets to an E3 ligase. Due in part to these reasons, protein degraders have been identified as new modalities in drug discovery and hold a promising future as research tools.
As protein degraders interact with both a target protein and E3 ligase simultaneously, their design is not trivial. A modular approach is often taken to generate PROTAC® libraries to screen for degradation of target proteins. To do this, strategic permutations of E3 ligands, target warheads, and linkers are synthesized with growing attention to linker properties and exit vectors. Each member of the target-specific degrader library is analyzed for depletion of the target in cells, providing leads for further optimization and characterization.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Visit our document search for data sheets, certificates and technical documentation.
Related Articles
- Protein Degrader Building Blocks are a collection of crosslinker-E3 ligand conjugates with a pendant functional group for covalent linkage to a target ligand.
- Targeted protein degradation reduces disease-relevant proteins in cells using small molecules, hijacking endogenous proteolysis systems.
- Solvias MeOBIPHEP Ligands: State-of-the-art atropisomeric MeOBIPHEP ligands, also referred to as MeO-BIPHEP, originally developed by Roche, have an extraordinarily broad performance profile for many synthetic applications due to their modular ligand design.
- Buchwald G6 precatalysts are oxidative addition complexes that are thermally, air, and moisture stable. The catalyst activation doesn’t require a base and generates innocuous byproducts.
- See All (11)
Related Protocols
- Highly characterized cryopreserved human colonic organoids and a step-by-step organoid culture protocol for epithelial intestinal organoid differentiation from iPS cells.
- Adhere cells to solid substrates using poly-lysine, which enhances electrostatic interaction between negatively charged ions of the cell membrane and the culture surface.
- This gelatin coating protocol for cell culture applications offers information regarding types of gelatin, surface coverage concentration, and tips for optimization.
- Coating surfaces with laminin for culturing cells requires specific conditions for optimal results. Protocols for coating coverslips to culture neurospheres and general cell culture are included.
- See All (0)
Find More Articles and Protocols
How Can We Help
In case of any questions, please submit a customer support request
or talk to our customer service team:
Email [email protected]
or call +1 (800) 244-1173
Additional Support
- Chromatogram Search
Use the Chromatogram Search to identify unknown compounds in your sample.
- Calculators & Apps
Web Toolbox - science research tools and resources for analytical chemistry, life science, chemical synthesis and materials science.
- Customer Support Request
Customer support including help with orders, products, accounts, and website technical issues.
- FAQ