MilliporeSigma
Reliable Results
Air and Fluid Particle Contamination Monitoring for Industrial Testing
Industrial testing is critical for preventive maintenance, QC analysis, and worker safety during the processing, manufacturing, packaging, and transportation of products. The levels of particle contamination tolerated in industrial processes are often extremely low and must be monitored. Fluids and fluid streams that are routinely monitored include fuels, gases, hydraulic oils, lubricants, water, chemicals, precision component rinses, cleaning tanks, boilers, and pharmaceutical products. Routine analysis of clean rooms, building abatements, and workplace environments relies on methodologies for particle analysis of air. National and international regulatory agencies have established standard methods of analysis and set threshold limits for a large number of liquid and airborne contaminants.
We specialize in manufacturing a variety of analytical tools for collecting, identifying, and measuring particles in air and fluids in manufacturing processes and industrial environments to meet today’s demanding standards.
Industrial Testing Supplies by Category
Aerosol Monitors and Cassettes
Three-piece 25- and 37-mm aerosol analysis monitors are available with 0.45 μm and 0.8 μm cellulose ester membranes. Matched-weight monitors contain two filters matched in weight to within 0.1 mg, eliminating test analysis. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface.
Aerosol Monitors, Complete with Membrane Filters
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Monitor Refills and Matched Weight Filter Pairs
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Empty Monitor Cassettes and Spare Rings
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Filter selection for air monitoring applications depends on the contaminant. Pore size, filter compatibility, and analytical method all play a part in filter choice. Our filters display high particle-collection efficiency over a broad range of particle sizes. We offer a variety of filter materials in various filter diameters and pore sizes to fit each specific application.
Cellulose (Mixed Cellulose Ester)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Glass Fiber
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Polycarbonate (PC)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Polyvinyl Chloride (PVC)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Polytetrafluoroethylene (PTFE)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Quartz Fiber
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Support Pads
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Orifices and Filter Holders
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
You can use different types of filters for fluid particle monitoring: membrane filters and depth filters.
  • The membrane filter is a uniform, continuous mesh of polymeric material with defined pore size. These filters are available in various materials. The material type you use depends on its compatibility with the fluid you need to sample.
  • Depth filters have a matrix of randomly oriented fibers pressed, wound, or bonded together into a random matrix of flow channels. Unlike membrane filters, depth filters have a nominal pore size, causing the filter to retain a range of particles during initial use. Due to the random orientation of fibers, depth filters do become saturated and eventually the pores become larger with use. Therefore, we recommend you use them for prefiltration or as a support pad in many of the monitoring procedures.
Factors such as the size of the particles, fluid/filter compatibility, and the filtration method determine which filter you should use.
MCE (Mixed Cellulose Ester)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Polytetrafluoroethylene (PTFE)
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Quartz Fiber
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Support Pads
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
We offer filter holders, chemical duty pumps, forceps, and Petri dishes for fluid monitoring.
  • Due to their inert nature and broad chemical resistance, borosilicate glass filter holders are commonly used for research and small-scale filtrations.
  • Stainless steel filter holders feature corrosion resistance, strength, and resistance to bacterial adherence. Due to these advantages, stainless steel filter holders are most commonly used in industrial applications requiring pressure or high-pressure filtration.
Filter Holders and Accessories
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Chemical Duty Pumps
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Forceps
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
Petri Dishes
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
The Fluid Sampling Kit allows sampling of liquids from pressurized systems through contamination analysis cassettes, for gravimetric or colorimetric determination of contaminant level. The stainless sampler assembly, containing a cassette, is plugged into a quick-release valve installed in the system line, and a measured quantity of liquid is allowed to pass through the cassette filter. Residual liquid is then removed from the cassette by using a valved syringe as a pump. For the gravimetric analysis, Matched-Weight Monitors (available as accessories) remove the need for pre-weighing filters before sampling.
Applications: Contamination analysis of aviation fuels, fluid storage tanks, cooling tanks.
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
The Patch Test Kit allows rapid monitoring of the degree of particulate contamination in hydrocarbon-based hydraulic fluids and lubricating oils. This is accomplished by observing the degree of discoloration of a 5 μm 47 mm disc filter through which a standard volume of the test liquid has been drawn by vacuum filtration. A membrane filter color rating scale and particle assessment scale corresponding to recognized standard levels of contamination is provided for comparison in determining observed contamination levels. Test results are dependably repeatable and sensitive enough to detect any significant change in cleanliness. Free water will appear either as droplets during the test procedure, or as a stain on the test filter. Filters are included for 100 tests.
Applications
Contamination analysis of hydrocarbon-based hydraulic fluids, lubricating oils, bulk chemicals.
  • Patch-testing hydraulic fluids
  • Contamination testing of lubricants
  • Purging fluid
  • Particulate contamination in boiler water
Sorry, an unexpected error has occurred
Response not successful: Received status code 500