MilliporeSigma
Laboratory Labels
Labels for laboratory use and sample management have special requirements based on sample type, container size, storage conditions, and application. Laboratory sample labeling presents many challenges such as solvent compatibility or the ability to remain adherent during freezing and cryostorage. Systems designed specifically for laboratory research labeling offer lab-specific symbols, auto serialization, and built in bar coding ensuring that you have the functionality for your application. The MilliSentials™ Lab Labeling System  provides a complete laboratory labeling solution with adaptable laboratory grade labels, a compact WIFI capable printer, and custom-designed laboratory labeling software. Specific labels such as  CRYO-BABIES®, CRYO TAGS® and Freezerbondz™ for cryostorage, and TOUGH-TAGS® that resist most aggressive chemicals can give you the confidence that your valuable samples can always be identified.
MilliSentials™ Lab Labeling System
  • The MilliSentials™ Lab Labeling System offers a complete solution for laboratory sample labeling. Lab grade labels are capable of withstanding temperatures from -196 °C (liquid nitrogen) to 100 °C (boiling water) and harsh chemical solvents (including ethanol, DMSO, xylene). The labels have also been designed with perforations allowing one size to fit across a wide range of laboratory vessels including tubes, cryovials, petri dishes, cell culture plates, and reagent bottles. The custom-built software features pre-constructed label templates for laboratory vessels, allows labels to be saved for quick repeat printing, and features a spreadsheet upload function for large batch printing. 
  • CRYO-BABIES®/CRYO-TAGS® Cryogenic labels can withstand temperatures from -196 °C  to  80 °C and are available in sizes to fit vials, microtubes, freezer boxes, microplates, cell culture plates, plastic bags, and other containers for storage in liquid nitrogen, freezers, and when working with samples on dry ice. 
  • TOUGH-TAGS® are chemically resistant labels that can withstand direct exposure or immersion in organic solvents, alcohols, and harsh or corrosive chemicals. TOUGH-TAGS® labels resist temperatures from -40° C to 121 °C  including boiling water baths and autoclaving making them useful for the identification of materials that require autoclave sterilization, and are manufactured with a high-performance adhesive for metal, glass, and plastic containers. 
  • Our printing solutions are designed for sample management and inventory tracking and can be integrated into your lab management workflow. 
  • A range of tapes are available for identifying samples, equipment, and materials in the lab, and are available in a variety of colors, sizes, and configurations for color coding and convenience. 
  • We also offer markers for a variety of laboratory applications, including ethanol- and alcohol-resistant markers, cryogenic markers, and water-resistant markers in an assortment of colors