Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Single transfection of a synthetic polycistronic self-replicative RNA yields high numbers of transgene-free human iPSCs
By: Min Lu1, Naohisa Yoshioka2, Steven Dowdy2 and Vi Chu1*,
1EMD Millipore, Bioscience Division, Temecula, CA, USA
2University of California, San Diego, La Jolla, CA, USA
Introduction
A ready source of induced pluripotent stem cells (iPSCs) is critical to the effective study of differentiation pathways and the investigation of the therapeutic potential of iPS cells. Since the discovery that human iPSCs could be generated by inducing expression of the four reprogramming factors (OCT-4, SOX-2, KLF-4 and c-MYC), many different reprogramming technologies have emerged to generate iPSCs, each possessing their own advantages and disadvantages. First-generation technologies, based on retroviral and lentiviral systems, allowed for highly efficient reprogramming events but lacked the necessary control over host genome integrations. Cre-excisable lentiviral systems offered a solution to genome integration but required lengthy subcloning procedures and screening to ensure excision of the reprogramming factors. Second-generation technologies used non-integrating episomal DNA plasmids, which were transgene-free but lacked the high reprogramming efficiencies of earlier retroviral and lentiviral techniques. Third-generation technologies used negative sense, non-integrating RNA viruses, termed Sendai Viruses (SeV), which originated from highly transmissible respiratory tract infections in mice, hamsters, guinea pigs, rats, and pigs. These RNA viruses produced integration-free iPSCs, produced high reprogramming efficiencies and were easy to use, but residual Sendai virus was difficult to clear from cells, resulting in the requirement for multiple rounds of clonal expansion and analysis.
The Simplicon™ RNA Reprogramming Technology is a next generation reprogramming system that uses a single synthetic, polycistronic self-replicating RNA strand engineered to mimic cellular RNA to generate human iPS cells. The single RNA strand contains the four reprogramming factors, OCT-4, KLF-4, SOX-2 and GLIS1, and enables extremely efficient reprogramming using a single transfection step without any viral intermediates or host genome integration. Once iPSCs are generated, the RNA can easily be selectively eliminated by removing the interferon-gamma (IFNg) inhibitor, B18R, from the cell culture medium.
Human iPS cells can be generated with a single transfection of polycistronic RNA
Figure 1. Human iPS cells can be generated with a single transfection of polycistronic RNA. Once created, the reprogramming RNA can be selectively degraded by removing B18R from the culture media, creating transgene-free, replicon-free iPSCs.
The Evolution of Reprogramming Technologies
Figure 2. The evolution of reprogramming technologies has culminated in the development of synthetic RNA-mediated reprogramming (extreme right), representing the safest and most efficient method for iPS cell generation: Simplicon™ RNA Reprogramming Technology combines the efficiency of retroviral and lentiviral reprogramming technologies with the safety of nonviral based reprogramming methods.
Methods
Human Reprogramming Timeline
Figure 3. Timeline showing points at which Simplicon™ reprogramming steps were performed.
Generation of human iPSCs using Simplicon™ RNA Technology
4 x 105 human foreskin fibroblasts were plated in each well of a 6-well plate in low serum fibroblast medium and allowed to attach overnight. HFFs were pretreated with B18R growth factor (200 ng/mL) for 2 h at 37 ˚C and 5% CO2. HFFs were then transfected with 1 μg of Simplicon™ VEE-OKS-iG and B18r RNA in 2.5 μL of RiboJuice™ mRNA transfection reagent following the manufacturer’s protocol. The mixture of Simplicon™ RNA and transfection reagent was incubated at 37 ˚C, 5% CO2 for 3 h. Following transfection with RNA, medium was exchanged with 2 mL/well of ADMEM medium containing 10% fetal bovine serum (FBS), 1% Gluta-MAX™ supplement and B18R protein (200 ng/mL).
Starting the day after transfection, cells were fed daily with ADMEM with 10% FBS, 1% GlutaMAX™ supplement, B18R protein and 0.5 μg/mL puromycin for a total of 10 days. 30-60% cell death was observed from day 4-5 and puromycin-resistant cells started to grow back at day 7-9 after puromycin selection.
At day 10, approximately 5 x 104 to 1 x 105 reprogrammed cells were replated on fresh EmbryoMax® Primary Mouse Embryo Fibroblasts in MEF-conditioned medium containing B18R protein (200 ng/mL) supplemented with small molecules contained in the Human iPS Reprogramming Boost Supplement II. Cell morphology was monitored daily, and small iPS cell colonies started to appear around day 15-16. At day 20, reprogrammed cells were transitioned to standard human embryonic stem cell medium, such as PluriSTEM Human ES/iPS media, without B18R protein and colonies were selected based on colony morphology and expanded for future experiments.
Results
Transfection efficiency using Simplicon™ RNA Reprogramming Technology
HFFs were transfected with either a control RNA replicon encoding green fluorescent protein (GFP) alone (Figure 4A) or with the Simplicon™ Reprogramming RNA (Figure 4B), and analyzed the following day to determine transfection efficiency. A high percentage of cells in Figure 4A showed GFP signal; transfection of HFFs using Simplicon™ Reprogramming RNA resulted in roughly 5-10% Oct-4 expression one day after transfection (green, Figure 4B).
Human Reprogramming Timeline
Figure 4. Fluorescent micrographs of HFFs transfected with a GFP RNA replicon control (A) or Simplicon™ Reprogramming RNA (B) and analyzed the following day to estimate transfection efficiency. The green signal in (B) reflects OCT-4 expression.
Reprogramming kinetics
To determine the kinetics of reprogramming, Simplicon™ transfected HFFs were analyzed at four timepoints by brightfield microscopy (Figure 5). Colonies of human iPSCs emerged between days 17-21 after a single transfection at day 1.
Human Reprogramming Timeline
Figure 5. Brightfield microscopy shows HFFs being reprogrammed over a 21 day time period using a single transfection of Simplicon™ Reprogramming RNA. After 10 days of puromycin selection and replating on fresh PMEF cells, colonies started to emerge at day 17-21.
Characterization of human iPSCs
Simplicon™ reprogrammed fibroblasts were assayed for alkaline phosphatase activity, a pluripotency marker, 28 days after Simplicon™ transfection (Figure 6). Both HFF and BJ fibroblasts showed high expression levels of alkaline phosphatase activity, with the HFF cells showing the most intense signal.
Alkaline phosphatase staining of day 28 reprogrammed fibroblasts (HFF and BJ) generated using Simplicon™ RNA Reprogramming Technology.
Figure 6. Alkaline phosphatase staining of day 28 reprogrammed fibroblasts (HFF and BJ) generated using Simplicon™ RNA Reprogramming Technology.
Lot-to-lot reproducibility
To compare the performance of different manufacturing lots of the synthetic Simplicon™ RNA strands, we used four different lots to transfect HFFs (10,000 cells per transfection) and then counted the number of hiPSC colonies that appeared 17-21 days later. Calculating reprogramming efficiency as the number of reprogrammed colonies divided by the number of original cells, we determined that all the manufacturing lots displayed similar reprogramming efficiencies, ranging from 0.5% to 1%.
Figure 7. Similar reprogramming efficiencies ranging from 0.6%-1% were generated using different manufacturing lots of Simplicon™ RNA.
Discussion
We have described a unique, synthetic, polycistronic RNA-based technology that enables efficient reprogramming of human somatic cells into induced pluripotent stem cells using a single transfection event. The Simplicon™ RNA reprogramming technology does not rely on lentiviral, retroviral or RNA-based viruses and is a step toward a more defined system for iPS cell generation. This RNA-based reprogramming approach has broad applicability for the efficient generation of hiPSCs for use in disease cell-modeling studies, transdifferentiation studies, and eventual human cell therapy/regenerative medicine applications.
Materials
 References
  1. Yamanaka S., et al. Induction of pluripotent stem cells from mouse embryonic and adult fibroblast cultures by defined factors. Cell. 2006 Aug 25;126(4):663-76.
  2. Ding, S, et al. Reprogramming of human primary somatic cells by OCT4 and chemical compounds. Cell Stem Cell. 2010 Dec 3;7(6):651-5.
  3. Dowdy, SF, et al. Efficient generation of human iPSCs by a synthetic self-replicative RNA. Cell Stem Cell. 2013 Aug 1;13(2):246-54.
Related Links