[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[C–H Functionalization](https://www.sigmaaldrich.com/US/en/applications/chemistry-and-synthesis/synthetic-methods/ch-functionalization)Diversinates™: Any-Stage Functionalization of (Hetero)aromatic Scaffolds # Diversinates™: Any-Stage Functionalization of (Hetero)aromatic Scaffolds ![Diversinates™ can functionalize heteroaromatic C–H bonds without the need for pre-functionalization, further saving time for the discovery chemist.](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/chemistry-and-synthesis/ch-functionalization/diversinate-ts2.jpg "Diversinates™ can functionalize heteroaromatic C–H bonds without the need for pre-functionalization, further saving time for the discovery chemist.") ## Introduction The synthesis of heteroaromatic and aromatic compounds is at the heart of the chemical industry. With the ever-growing demand of new chemical entities coupled to the dwindling resources and time constraints allotted to any given research project, a rapid way to diversify (hetero)aromatic scaffolds is needed. With operational simplicity (reactions that are not sensitive to air and moisture) in mind, a series of reagents that can append a variety of useful functionalities onto (hetero)aromatic cores was designed. Diversinates™ can functionalize heteroaromatic C–H bonds without the need for pre-functionalization, further saving time for the discovery chemist. ## Advantages Although metal-catalyzed cross-coupling chemistry is robust and undeniably useful, it relies on pre-functionalized (often halogenated) starting materials. Metal-catalyzed C–H activation is of recent research interest; and achieving the (fluoro)alkylation of heteroarene scaffolds in this manner is still a challenge. The Baran Diversinates™ can react with (hetero)arene cores that do not have apparent chemical handles—only heteroaromatic C–H bonds are needed. These powerful reagents take advantage of the innate reactivity of the substrate and react under practical reaction conditions (without the need to exclude air and moisture). ## Representative Applications ![Baran Diversinates](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/chemistry-and-synthesis/ch-functionalization/rxn-scheme-diversinates-ts.jpg "Baran Diversinates") __Figure 1.__Representative Applications ## Materials Sorry, an unexpected error has occurred Response not successful: Received status code 500 ### References 1\. Fujiwara Y, Dixon JA, Rodriguez RA, Baxter RD, Dixon DD, Collins MR, Blackmond DG, Baran PS. 2012. A New Reagent for Direct Difluoromethylation. J. Am. Chem. Soc.. 134(3):1494-1497. [https://doi.org/10.1021/ja211422g](https://doi.org/10.1021/ja211422g) 2\. Fujiwara Y, Dixon JA, O?Hara F, Funder ED, Dixon DD, Rodriguez RA, Baxter RD, Herlé B, Sach N, Collins MR, et al. 2012. Practical and innate carbon?hydrogen functionalization of heterocycles. Nature. 492(7427):95-99. [https://doi.org/10.1038/nature11680](https://doi.org/10.1038/nature11680) 3\. Zhou Q, Ruffoni A, Gianatassio R, Fujiwara Y, Sella E, Shabat D, Baran PS. 2013. Direct Synthesis of Fluorinated Heteroarylether Bioisosteres. Angew. Chem. Int. Ed.. 52(14):3949-3952. [https://doi.org/10.1002/anie.201300763](https://doi.org/10.1002/anie.201300763) 4\. Gui J, Zhou Q, Pan C, Yabe Y, Burns AC, Collins MR, Ornelas MA, Ishihara Y, Baran PS. 2014. C?H Methylation of Heteroarenes Inspired by Radical SAM Methyl Transferase. J. Am. Chem. Soc.. 136(13):4853-4856. [https://doi.org/10.1021/ja5007838](https://doi.org/10.1021/ja5007838) 5\. Gianatassio R, Kawamura S, Eprile CL, Foo K, Ge J, Burns AC, Collins MR, Baran PS. 2014. Simple Sulfinate Synthesis Enables C?H Trifluoromethylcyclopropanation. Angew. Chem. Int. Ed.. 53(37):9851-9855. [https://doi.org/10.1002/anie.201406622](https://doi.org/10.1002/anie.201406622) __Related Articles__ - [C–H Amination](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/c-h-amination) - [Jørgensen’s Organocatalysts](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/jorgensen-organocatalysts) - [MCAT- 53™ Catalyst for Ruthenium Formation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/mcat-53-green-ruthenium-catalyst) - [Palau’Chlor®: Easily and Selectively Functionalize Lead Compounds](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/palauchlor) - [Role of High Purity Metal Salts in Chemical Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/role-of-high-purity-metal-salts-in-chemical-synthesis) - [Ellman's Sulfinamides](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/sulfinamids) - [SuFEx: Sulfonyl Fluorides that Participate in the Next Click Reaction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/sulfonyl-fluorides) - [Electrochemical Allylic C–H Oxidation](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/synthetic-reagents-electrochemical-allylic-oxidation-tcnhpi) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) __Related Product Categories__ - [C–H Activation Catalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/c-h-activation-catalysts) - [Boronic Acids & Derivatives](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-synthesis/boronic-acids-and-derivatives) - [Environmental Accounting Supplier Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/environmental-accounting-supplier-toolkit) - [Renewable Electricity Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/renewable-electricity-toolkit) - [Crown Ethers and Cryptands for Advanced Applications](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/crown-ethers-cryptands-advanced-applications) - [Purification of Extracellular Vesicles with Eshmuno® Q Resin and Natrix® Q Membrane](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/membrane-chromatography/purification-extracellular-vesicles) - [Sterilizing filters for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/final-sterile-filtration-and-filling/sterilizing-grade-membrane-filters) - [Single-use assemblies for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/mobius-single-use-assemblies/single-use-filtration-assemblies) - [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search) Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map) An unknown error has occured.